Gel Knife
Invitrogen™

Gel Knife

Replacement Parts for Electrophoresis ApparatusRead more
Have Questions?
Catalog NumberQuantity
EI90101 Each
Catalog number EI9010
Price (USD)
77.65
Online exclusive
80.50
Save 2.85
Each
Quantity:
1 Each
Price (USD)
77.65
Online exclusive
80.50
Save 2.85
Each
Replacement Parts for Electrophoresis Apparatus
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Each
Shipping ConditionRoom Temperature
For Use With (Application)1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
For Use With (Equipment)Universal
Gel SizeMini
Product LineNovex, XCell SureLock
TypeCassette Opener
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

Can I purchase replacement parts for the Mini Blot Module?

Here are the replacement parts we offer for the Mini Blot Module:

- Mini Blot Module Gasket, Cat. No. B1001
- Gel Knife, Cat. No. EI9010
- Bolt Tank Base, Cat. No. B4478640
- Gel Runner Tank, Cat. No. B4478641
- Mini Gel Tank Lid, Cat. No. A25944
- Cassette Clamp, Right, Cat. No. B4478592
- Cassette Clamp, Left, Cat. No. B4478593
- Power Supply Adapters, Cat. No. ZA10001

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What are the dimensions of the Gel Knife?

The dimensions of the Gel Knife are: Handle 13 cm, Blade (9 cm (length) x 4.5 cm (width).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Can I purchase replacement parts for the XCell SureLock Mini-Cell?

Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What are the components of the XCell SureLock Mini-Cell?

The XCell SureLock Mini-Cell includes the lid, gel tension wedge, lower buffer chamber, buffer core, buffer dam, and gel knife.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.