Kit de detección colorimétrica de glucosa
Invitrogen™

Kit de detección colorimétrica de glucosa

The Glucose Detection research-use-only kit is a colorimetric assay designed for the quantification and detection of glucose in serum, plasma,Más información
Have Questions?
Número de catálogoCantidad
EIAGLUC2 × 96 pruebas
Número de catálogo EIAGLUC
Precio (MXN)
-
Cantidad:
2 × 96 pruebas
Pedido a granel o personalizado
The Glucose Detection research-use-only kit is a colorimetric assay designed for the quantification and detection of glucose in serum, plasma, urine, buffers and tissue culture media.

This complete, ready-to-use kit includes clear 96-well plate(s), glucose standard, assay buffer, and other components to perform the assay. A 96-well microplate reader capable of reading optical density at 560 nm is required for use of this kit.

Performance characteristics
• Assay type: colorimetric detection kit
• Sample types: serum, plasma, urine, buffers, and tissue culture media
• Sensitivity: 0.413 mg/dL
• Standard curve range: 0.5–32 mg/dL
• Reactivity: species independent

Background
Glucose is by far the most common carbohydrate. It is a monosaccharide, an aldose, a hexose, and a reducing sugar and is also known as dextrose, because it is dextrorotatory (rotates polarized light clockwise). The structure of glucose is shown below as both the straight chain and cyclic forms. For all biological and molecular events and for multiple cellular functions, energy is essential.

Energy is available in the form of ATP (adenosine triphosphate), most of which is generated through aerobic cellular respiration of carbohydrate and glucose, the major source of biological free energy in higher organisms. Reduced energy levels threaten cellular homeostasis and integrity. Impaired energy metabolism may trigger pro-apoptotic signaling (programmed cell death), oxidative damage, excitotoxicity, and impede mitochondrial DNA repair.

Assay principle
The Glucose Colorimetric Detection Kit is designed to quantitatively measure glucose in a variety of samples. A beta-D-glucose standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Samples are mixed with the colorimetric substrate and horseradish peroxidase and the reaction initiated by addition of glucose oxidase. The reaction is incubated at room temperature for 30 minutes. The glucose oxidase reacts with glucose to produce hydrogen peroxide which, in the presence of HRP, reacts with the colorimetric substrate to convert the colorless substrate into a pink-colored product. The pink product is read at 560 nm. Increasing levels of glucose cause a linear increase in color.

Related links
Learn more about ELISA kits
Learn more about other immunoassays

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Para utilizar con (equipo)Lector de microplacas
ID de gen (Entrez)Sin genes
Símbolo de genSin genes
Tipo de productoKit de detección colorimétrica de glucosa
Cantidad2 × 96 pruebas
Condiciones de envíoAprobado para su envío en hielo húmedo o seco
Nombre del kit de destinoGlucosa
Método de detecciónColorimétrico
Formato del kit de inmunoensayoVetMAX™-Gold AIV
Tipo de muestraotros fluidos biológicos, plasma, Suero
Unit SizeEach
Contenido y almacenamiento
Placas de 96 pocillos, estándar, otros reactivos específicos del ensayo, tampones y hoja de datos técnicos específicos del lote.

Almacenar a –20 °C.

Preguntas frecuentes

Can I use the Glucose Colorimetric Detection Kit (Cat. No. EIAGLUC) with mouse samples?

The Glucose Colorimetric Detection Kit (Cat. No. EIAGLUC) is designed to measure glucose in various sample types, including serum, plasma, urine, buffered solutions, and tissue culture medium. While the assay was characterized with human samples, the kit is described as species-independent, meaning it should be suitable for use with samples from other species including mouse samples.

However, it is always recommended to validate the assay with your specific sample type to help ensure accurate and reliable results.