Heratherm Incubation Chambers
Heratherm Incubation Chambers
Heratherm Incubation Chambers
Heratherm Incubation Chambers
Heratherm Incubation Chambers
Thermo Scientific™

Heratherm Incubation Chambers

Comprehensive data monitoring and temperature control combined with a flexible Heratherm Incubation Chamber provide a dependable option for microbiological incubation or temperature-dependent storage and testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDoor StyleCapacity (Metric)
EIG250Glass door247 L
EIS750Solid (outer), Glass (inner)750 L
EIG750Glass750 L
EIS250Solid outer door with inner glass door247 L
Catalog number EIG250
Price (USD)
-
Door Style:
Glass door
Capacity (Metric):
247 L
Produces precise temperature conditions above and below ambient, ideally suited for microbiological incubation, and heated or cooled incubated storage.
  • Uniform Temperature throughout the chamber.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
Specifications
Alarm TypesAudible and Visible
Capacity (English)8.8 cu.ft.
Capacity (English) Shelf66.1 lb.
Capacity (Metric) Shelf30 kg
Certifications/ComplianceUL, CE
Data OutputsUSB, Thermo Cloud via OPC UA
Depth (English)31.1 in.
Depth (English) Interior20.4 in.
Depth (Metric)790 mm
Depth (Metric) Interior519 mm
Display7 in. Touchscreen user interface
Door StyleGlass door
Electrical Requirements100/240V, 50/60 Hz
For Use With (Application)Incubation, storage, temperature testing
Height (English)44.4 in.
Height (English) Interior31.5 in.
Height (Metric)1128 mm
Height (Metric) Interior800 mm
LightingNo
No. of CastersNone
Plug TypeNema 5-20P
Ports2 access ports
ProgrammabilityTemperature ramping
Shelf Area (English)3.05 ft.2
Shelf Area (Metric)0.28 m2
Shelf MaterialStainless Steel (304/1.4301)
Shipping Weight (English)448 lb.
Shipping Weight (Metric)203 kg
Temperature ControlMicroprocessor - programmable
Temperature Resolution0.1°C
Temperature Stability≤ ±0.1°C @ 20°C to 37°C
Temperature Uniformity≤ ±0.5°C @ 20°C to 37°C
TypeTemperature Test Chamber
Warranty24 months
Wattage600 W
Width (English)35.5 in.
Width (English) Interior26 in.
Width (English) Shelf24.2 in.
Width (Metric)901 mm
Width (Metric) Interior660 mm
Width (Metric) Shelf615 mm
Capacity (Metric)247 L
Chamber MaterialPowder-coated Cold Rolled Steel
MaterialPowder-coated Cold Rolled Steel
No. of Shelves2
Relative Humidity10 to 90%
Temperature Range (Metric)0 to 70°C
Voltage100/115/220/230 V
Unit SizeEach