Heratherm Incubation Chambers
Thermo Scientific™

Heratherm Incubation Chambers

Comprehensive data monitoring and temperature control combined with a flexible Heratherm Incubation Chamber provide a dependable option for microbiological incubation or temperature-dependent storage and testing.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoModelo de puertaCapacidad (métrico)
EIG750Vidrio750 L
EIS750Sólido (exterior), vidrio (interior)750 L
EIS250Solid outer door with inner glass door247 L
EIG250Glass door247 L
Número de catálogo EIG750
Precio (USD)
-
Modelo de puerta:
Vidrio
Capacidad (métrico):
750 L
Produces precise temperature conditions above and below ambient, ideally suited for microbiological incubation, and heated or cooled incubated storage.
  • Uniform Temperature throughout the chamber.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
Especificaciones
Tipos de alarmaAudible and Visible
Capacidad (imperial)26.5 cu. ft.
Capacidad del estante (imperial)66.1 lbs
Capacidad del estante (métrico)30 kg
Certificaciones/conformidadUL, CE
Salidas de datosUSB, Ethernet
Fondo (imperial)34.84 in.
Fondo interior (imperial)24.8 in.
Fondo (métrico)885 mm
Fondo interior (métrico)630 mm
Pantalla7 in. touchscreen user interface
Modelo de puertaVidrio
Requisitos eléctricos100/240V, 50/60 Hz, 1400W
Altura (imperial)72.24 in.
Altura (imperial) interior57.09 in
Altura (métrico)1835 mm
Altura (métrico) interior1450 mm
IluminaciónNo
N.º de ruedas giratorias4
Tipo de tapónNema 5-20P
Puertos2 access ports
ProgramableTemperature ramping
Área del estante (imperial)4.9 ft²
Área del estante (métrico)0.46 m²
Material del estanteStainless Steel (304/1.4301)
Peso con embalaje (imperial)820 lbs
Peso con embalaje (métrico)372 kg
Control de temperaturaMicroprocessor - programmable
Resolución de temperatura0,1 °C
Estabilidad de la temperatura≤ ±0.1°C @ 20°C to 37°C
Uniformidad de temperatura≤ ±0.5°C @ 20°C to 37°C
TipoCámara climática
Garantía24 months
Vataje900 W
Anchura (imperial)42.52 in.
Anchura (imperial) interior33.07 in.
Anchura (imperial) del estante22.6 in.
Anchura (métrico)1080 mm
Anchura (métrico) interior840 mm
Anchura (métrico) del estante795 mm
Capacidad (métrico)750 L
Material de la cámaraPowder-coated Cold Rolled Steel
MaterialPowder-coated Cold Rolled Steel
N.º de estantes3
Intervalo de temperatura (métrico)0 to 70°C
Tensión100/115/220/230 V
Unit SizeEach