Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Thermo Scientific™

Heratherm Light Chambers 750 L

Accurate and even light distribution with Long-lasting LED lighting for (ICH) light testing, plant growth and animal hatching — relative humidity and temperature are controlled to meet specified environment.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoModelo de puertaPara utilizar con (aplicación)Incluye
ELS750Puerta maciza con puerta interior de cristalPruebas de luz ICH, crecimiento de plantas, pruebas de luz, eclosión de animalesMódulos de luz no incluidos - Prueba ICH: 333979G01 / Estándar: 333980G01
ELG750Puerta de vidrioCrecimiento de plantas, pruebas de luz, eclosión de animalesMódulos de luz no incluidos - para crecimiento de plantas/pruebas/eclosión de animales: 333980G01
Número de catálogo ELS750
Precio (MXN)
-
Modelo de puerta:
Puerta maciza con puerta interior de cristal
Para utilizar con (aplicación):
Pruebas de luz ICH, crecimiento de plantas, pruebas de luz, eclosión de animales
Incluye:
Módulos de luz no incluidos - Prueba ICH: 333979G01 / Estándar: 333980G01
Accurate control of three parameters - Temperature above and below ambient, relative humidity and light - provide great environment in testing applications and for biological research with plants and animals. Choice of ICH module provides capability for efficient ICH light testing in Pharma.
  • Uniform Temperature / relative humidity / light exposure for samples.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • LED light system provides specified light exposure on 1, 2 or 3 shelves; programmable to adapt light quality / intensity and on/off timing - e.g. for day-night cycles. Choose from two light modules (ordered separately):
    • 1. ICH module, includes UV LED for ICH light testing;can only be used in ELS750 - model with solid outer door - for safety reasons
    • 2. Testing module for plant growth / daylight testing / animal hatching
  • Environmentally friendly water-efficient humidification system - with water tank or optional fix water connection for in-house purified water
  • Intuitive touchscreen interface with easy programmability of parameter cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard (with Connect Edge solution)
  • Sample and process protection: Assign personalized access with passcode; restrict access with standard key door lock or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Exterior glass door also available (not for ICH module use - for safety reasons).
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
Especificaciones
Tipos de alarmaAudible and Visible
Capacidad (imperial)26.5 cu.ft
Capacidad del estante (imperial)66 lbs (stationary)
Capacidad del estante (métrico)30 kg (stationary)
Certificaciones/conformidadUL, CE
Salidas de datosUSB, Thermo Cloud via OPC UA
Dimensiones (Largo x Ancho x Alto)895 x 1080 x 1835 mm / 35.24 x 42.52 x 72.24 in
Dimensiones (L x An x Al) Interior630 x 840 x 1450 mm / 24.8 x 33.07 x 57.09 in
Pantalla7 in. Touchscreen user interface
Modelo de puertaPuerta maciza con puerta interior de cristal
Requisitos eléctricos100 - 240V, 50-60Hz, 1700W
Para utilizar con (aplicación)Pruebas de luz ICH, crecimiento de plantas, pruebas de luz, eclosión de animales
Rango de humedad10 - 80% RH with light; 10 -90% RH w/o light
Resolución de humedad0.1% RH
Fuente de humedadRemovable water tank
IncluyeMódulos de luz no incluidos - Prueba ICH: 333979G01 / Estándar: 333980G01
IluminaciónLED
Tipo de tapónNema 5-20P
Puertos2 access ports
ProgramableTemperature, relative humidity, light intensity
Área del estante (imperial)4.9 ft2
Área del estante (métrico)0.46 m2
Material del estanteStainless Steel (304/1.4301)
Peso con embalaje (imperial)862 lbs
Peso con embalaje (métrico)391 kg
Resolución de temperatura0.1°C
Estabilidad de la temperatura≤±0.3°C @ 25°C to 40°C
Uniformidad de temperatura≤±2.0°C @ 25°C to 40°C
TipoVertical Light Chamber
Garantía24 months
Consumo de agua0.2 - 1.5 L per day; Value depends on settings, usage (e.g. number of door openings), and environmental conditions
Resistividad del agua0.05 - 1 MΩ
Vataje1700 W
Anchura (imperial) del estante31.3 in.
Anchura (métrico) del estante795 mm
Capacidad (métrico)750 L
Material de la cámaraStainless Steel (304/1.4301)
MaterialPowder-coated Galvanized Steel
N.º de estantes3
Humedad relativa10 to 90%
Intervalo de temperatura (métrico)15 to 45°C with light; 0 to 70°C w/o light
Tensión100/240 V
Unit SizeEach