This product is only available through your local Thermo Scientific supplier. Learn where to buy >

IL-10 Mouse ELISA Kit

Catalog number: EM2IL10

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Assay IL-10 (mouse interleukin-10) levels in mouse serum and culture supernatant samples with our Thermo Scientific Mouse IL-10 ELISA Kit.

This mouse IL-10 ELISA kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-10 (IL-10) in samples at 50 µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kit contains a pre-coated 96-well strip plate and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

Features of the Mouse IL-10 ELISA Kit:

Target: natural and recombinant mouse interleukin-10 (IL-10)
Format: colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: absorbance at 450nm minus absorbance at 550nm
Assay Range: 37 to 3000 pg/mL
Sensitivity: < 12 pg/mL
Precision: intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: cell culture supernatant, serum
Total Assay Time: 5 hours
Sample Size: 50 µL
Specificity: no significant cross-reaction with mouse IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, GM-CSF, IFNγ, TNFα or human IL-10.

Includes:
Complete kit contains a pre-coated antibody-plate, detection antibodies, buffers, diluents, standards, and substrates.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Cytokines & Receptors
Assay Range: See product documentation
Assay Sensitivity: See product documentation
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Pierce
Gene Aliases: CSIF, IL-10
Gene ID: 16153
Gene Name: interleukin 10
Gene Symbol: IL10
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 96 tests
Protein Aliases: IL-10, Cytokine synthesis inhibitory factor, CSIF
Protein Name: Interleukin-10
Target (Kit Named): IL-10
Target Species (Named): Mouse
Target Species (Validated): Mouse
UniProt ID: P18893

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols