Mouse 4-1BB (CD137/TNFRSF9) ELISA Kit - FAQs
Mouse 4-1BB (CD137/TNFRSF9) ELISA Kit - FAQs
View additional product information for Mouse 4-1BB (CD137/TNFRSF9) ELISA Kit - FAQs (EMTNFRSF9X10, EMTNFRSF9, EMTNFRSF9X5)