Search
View additional product information for RNase T1 (1000 U/μL) - FAQs (EN0541, EN0542)
1 product FAQs found
We would recommend 0.2 µL RNase T1 per µg of RNA in 50 mM Tris-HCl (pH 7.5) and 1 mM EDTA. Incubate at 37 degrees C for 30 min.