TheraPure™ GMP RNase Inhibitor, 40 U/μL, 400 kU - FAQs
TheraPure™ GMP RNase Inhibitor, 40 U/μL, 400 kU - FAQs
View additional product information for TheraPure™ GMP RNase Inhibitor, 40 U/μL - FAQs (EO038SKB007, EO038AFSKB001, EO038SKB011, EO038AFB036)
1 product FAQs found
What is the concentration of the TheraPure GMP RNase Inhibitor, 40 U/μL (Cat. No. EO038AFB036, EO038SKB011, EO038AFSKB001, EO038SKB007) in mg/mL, or the unit activity per mg of protein?
The concentration and activity of the TheraPure GMP RNase Inhibitor can be found in the Certificate of Analysis (CoA) provided with the product. Each lot may have different results, but the CoA will provide the activity range requirements.