DreamTaq DNA Polymerase (5 U/μL)
Thermo Scientific™

DreamTaq DNA Polymerase (5 U/μL)

Thermo Scientific DreamTaq DNA Polymerase is an enhanced Taq DNA polymerase optimized for all standard PCR applications. It ensures higher sensitivity, longer PCR products, and higher yields compared to conventional Taq DNA polymerase.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorQuantity
EP0712Green500 Units
EP0701Colorless200 Units
EP0702Colorless500 Units
EP0703Colorless5 x 500 Units
EP0705Colorless10 x 500 Units
EP0704Colorless20 x 500 Units
EP0711Green200 Units
EP0713Green5 x 500 Units
EP0714Green20 x 500 Units
Catalog number EP0712
Price (USD)
75.65
Online Exclusive
104.00
Save 28.35 (27%)
Each
Color:
Green
Quantity:
500 Units
Request bulk or custom format
Price (USD)
75.65
Online Exclusive
104.00
Save 28.35 (27%)
Each
Thermo Scientific DreamTaq DNA Polymerase is an enhanced Taq DNA polymerase optimized for all standard PCR applications. It ensures higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase. DreamTaq DNA Polymerase uses the same reaction setup and cycling conditions as conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required. It is supplied with optimized DreamTaq Buffer, which includes 20 mM MgCl2

DreamTaq Green DNA Polymerase is a combination of DreamTaq DNA Polymerase and 10X DreamTaq Green Buffer. The 10X DreamTaq Green Buffer includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with PCR performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Features

  • Robust amplification with minimal optimization
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Incorporates modified nucleotides, but does not incorporate dUTP
  • Direct loading of PCR product on gel with green format

Applications

  • Routine PCR amplification of DNA fragments up to 6 kb
  • High throughput PCR
  • Genotyping

Notes

  • For applications requiring PCR product analysis by absorbance or fluorescence excitation, colorless 10X DreamTaq Buffer (Cat. No. B65) is recommended for purifying PCR products before analysis.
For Research Use Only. Not for use in diagnostic procedures.
In certain countries use of this product is covered by patents. Purchase of product in these countries includes non-transferable, limited license for using only this amount of product for the purchaser's own internal research.

This product is licensed under one or more U.S. Patents Nos. 5,500,363 and 5,352,778 or corresponding foreign patents. This product is licensed under one U.S. Patent No. 5,436,149 or corresponding foreign patents owned by Takara Bio, Inc.

Patent pending.

Specifications
ColorGreen
Fidelity (vs. Taq)1X
Hot StartNo
Overhang3'-A
PolymeraseDreamTaq DNA Polymerase
Quantity500 Units
Reaction FormatSeparate Components
Concentration5 U/μL
For Use With (Application)Standard PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• DreamTaq DNA Polymerase (5 U/μL), 100 μL
• 10X DreamTaq Green Buffer with 20 mM MgCl2, 2 x 1.25 mL

Store at -20°C.

Frequently asked questions (FAQs)

What is the error rate of DreamTaq DNA Polymerases?

The error rate of DreamTaq DNA Polymerase is the same as that of Taq DNA Polymerase: 2.2x10^-5 errors per nt per cycle.