Search
Search

| Catalog Number | Color | Quantity |
|---|---|---|
| EP0714 | Green | 20 x 500 Units |
| EP0701 | Colorless | 200 Units |
| EP0702 | Colorless | 500 Units |
| EP0703 | Colorless | 5 x 500 Units |
| EP0705 | Colorless | 10 x 500 Units |
| EP0704 | Colorless | 20 x 500 Units |
| EP0711 | Green | 200 Units |
| EP0712 | Green | 500 Units |
| EP0713 | Green | 5 x 500 Units |
DreamTaq Green DNA Polymerase is a combination of DreamTaq DNA Polymerase and 10X DreamTaq Green Buffer. The 10X DreamTaq Green Buffer includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with PCR performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.
Features
Applications:
Notes
This product is licensed under one or more U.S. Patents Nos. 5,500,363 and 5,352,778 or corresponding foreign patents. This product is licensed under one U.S. Patent No. 5,436,149 or corresponding foreign patents owned by Takara Bio, Inc.
Patent pending.