Search
Search

| Catalog Number | Color | Quantity |
|---|---|---|
| EP1701 | Colorless | 200 U |
| EP1702 | Colorless | 500 U |
| EP1703 | Colorless | 2500 U |
| EP1711 | Green | 200 U |
| EP1712 | Green | 500 U |
| EP1713 | Green | 2500 U |
DreamTaq Hot Start DNA Polymerase employs antibody-based inhibition of DNA polymerase activity at ambient temperatures to prevent the amplification of non-specific products prior to the amplification step. With DreamTaq Hot Start DNA Polymerase, reactions can be set up at room temperature using the same protocol and cycling conditions as conventional Taq DNA polymerases. The enzyme is supplied with an optimized 10X DreamTaq Buffer containing magnesium chloride, which eliminates the need for extensive optimization of reaction conditions. DreamTaq Hot Start DNA Polymerase is also available with pre-added density reagent and electrophoresis tracking dyes, as well as in master mix format.
The 10X DreamTaq Green Buffer also includes a density reagent and two tracking dyes for convenient direct loading of PCR products on a gel.
DreamTaq Hot Start DNA Polymerase is formulated to enhance productivity through:
Applications: