Universal Assay Buffer (1X)
Invitrogen™

Universal Assay Buffer (1X)

Universal Assay Buffer
Have Questions?
Catalog NumberQuantity
EPX-11111-0001 each
Catalog number EPX-11111-000
Price (USD)
87.65
Online exclusive
101.00
Save 13.35
Each
Quantity:
1 each
Price (USD)
87.65
Online exclusive
101.00
Save 13.35
Each
The ProcartaPlex Universal Assay Buffer (1X) is developed for use with the ProcartaPlex assays. The ProcartaPlex Universal Assay Buffer (1X) is used for dilution of serum and plasma samples.
The ProcartaPlex Universal Assay Buffer (1X) is included in our basic kits and off the shelf panels. Additionally, we offer the Universal Assay Buffer as a stand-alone item.

Reported Application
Multiplex Immunoassay

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Luminex™ Instruments
Product TypeBuffer
Quantity1 each
SpeciesCanine, Mouse, Rat, Porcine, Non-Human Primate, Human
Unit SizeEach

Frequently asked questions (FAQs)

Why is it that the ProcartaPlex Universal Assay Buffer is needed for plasma/serum samples in ProcartaPlex assays?

The ProcartaPlex Universal Assay Buffer is needed to avoid matrix interference of serum and plasma with ProcartaPlex assay kit components.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Are the standalone ProcartaPlex assay buffers lot-specific such that they will only work with specific lots of other components?

No, the standalone ProcartaPlex assay buffers are not lot-specific.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I get the catalog numbers for ProcartaPlex assay buffers and plates?

Here are the catalog numbers:

- EPX-11111-000: ProcartaPlex Universal Assay Buffer (1X) (10 mL) - It replaces all discontinued Procarta buffers (standard, assay, sample buffer); it is the same as the discontinued Procarta Cell Lysate Assay Buffer, 10 plate (Cat. No. PC0165), but only for one plate
- EPX-11110-000: ProcartaPlex Universal Assay Buffer (10X) (10 mL)
- EPXP-11112-000: ProcartaPlex Platinum Assay Buffer for Plasma Samples (1X, 10 mL)
- EPXP-11113-000: ProcartaPlex Platinum Assay Buffer for Serum Samples (1X, 10 mL)
- EPX-99999-000: ProcartaPlex Cell Lysis Buffer; It is the same as the discontinued Procarta Cell lysis Buffer (for cell lysate preparation) (Cat. No. PC6002)
- EPX-22222-000: ProcartaPlex Detection Antibody Diluent, Mouse (1X, 3 mL)
- EPX-33333-000: ProcartaPlex Detection Antibody Diluent, Human (1X, 3 mL)
- EPX-66666-000: ProcartaPlex Wash Buffer 10X (20 mL)
- EPX-66666-001: ProcartaPlex Wash Buffer 10X (200 mL)
- EPX-77777-000: ProcartaPlex Reading Buffer (1X, 40 mL)
- EPX-SAPE-000: ProcartaPlex Streptavidin-PE (1X, 5 mL) - 4.3 µg/mL (can range from 4.25-4.35 µg/mL)
- EPX-44444-000: ProcartaPlex 96 well Flat Bottom Plate, black (1 plate)
- EPX-88888-000: ProcartaPlex 96 well Filter Plate (1 plate)
- EPX-38488-000: ProcartaPlex 384 well Filter Plate (1 plate)

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.