ProcartaPlex™ Cell Lysate Assay Diluent (1X)
Invitrogen™
ProcartaPlex™ Cell Lysate Assay Diluent (1X)
ProcartaPlex Cell Lysate Assay Diluent (1X) is for use with the Apoptosis 6-Plex Human ProcartaPlex Panel for dilution of cell lysate samples.
Have Questions?
Catalog NumberQuantity
EPX-11114-00010 mL
Catalog number EPX-11114-000
Price (USD)
197.00
Each
Quantity:
10 mL
Price (USD)
197.00
Each
ProcartaPlex Cell Lysate Assay Diluent (1X) is for use with the Apoptosis 6-Plex Human ProcartaPlex Panel (Cat. No. EPX060-15823-901) for dilution of cell lysate samples. Diluent is included with the panel, but is also offered here as a stand-alone item.
Reported application
  • Multiplex immunoassay
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineProcartaPlex
Shipping ConditionWet Ice
Product TypeDiluent
Quantity10 mL
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).