ProcartaPlex™ Human Viral Reagent Kit
Invitrogen™

ProcartaPlex™ Human Viral Reagent Kit

The ProcartaPlex Viral Reagent Kit is designed to be used with the Coronavirus Simplex kits to combine single analytes intoRead more
Have Questions?
Catalog NumberQuantity
EPX010-10430-90196 Tests
Catalog number EPX010-10430-901
Price (USD)
-
Quantity:
96 Tests

The ProcartaPlex Viral Reagent Kit is designed to be used with the Coronavirus Simplex kits to combine single analytes into a multiplex assay. This provides maximum flexibility to design assay panels as needed. This kit includes all reagents except for the target-specific beads (sold separately) to run the assay.

ProcartaPlex immunoassays are an ideal solution for assessing multiple protein biomarkers in a single sample.

Suitable sample types: Serum, Plasma

Sample volume: Serum, Plasma: 25 μL

Reported application: Multiplex Immunoassay

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay SensitivitySee Certificate of Analysis
For Use With (Equipment)Luminex Instruments
Product LineProcartaPlex
Shipping ConditionWet Ice
CombinabilityCombinable
Product TypeBasic and Setup Kits
Quantity96 Tests
SpeciesHuman
Unit SizeEach
Contents & Storage

• 1 vial Biotinylated Detection Antibody Mix (50X)
• 2 vials Human Control-High
• 2 vials Human Control-Medium
• 2 vials Human Control-Low
• 1 bottle Reading Buffer (1X)
• 1 bottle Wash Buffer (10X)
• 1 bottle Assay Buffer (1X)
• 1 bottle Detection Antibody Diluent (1X)
• 8-tube strip
• Adhesive film
• Flat bottom 96-well plate, black

Store at 2°C to 8°C.