ProcartaPlex™ Human Beta Endorphin Competitive Simplex Kit
Invitrogen™

ProcartaPlex™ Human Beta Endorphin Competitive Simplex Kit

The ProcartaPlex Human Beta Endorphin Competitive Simplex Kit measures Beta Endorphin protein and is based on competitive ELISA technique, where sample Testosterone competes with a labeled tracer for antibody binding, resulting in an inverse signal proportional to cortisol concentration.
Have Questions?
Catalog NumberQuantity
EPX010-12372-90196 Tests
Catalog number EPX010-12372-901
Price (USD)
351.00
Each
Quantity:
96 Tests
Price (USD)
351.00
Each

The ProcartaPlex Human Beta Endorphin Competitive Simplex Kit measures Beta Endorphin protein and is based on competitive ELISA technique, where sample Testosterone competes with a labeled tracer for antibody binding, resulting in an inverse signal proportional to cortisol concentration.

This kit can be used for single-target quantification or combined with other simplex assays to create a panel. ProcartaPlex Competitive Simplex kits are compatible with each other and can also be combined with sandwich assays. Please use the Panel Configurator to verify the compatibility of your desired panel.

A ProcartaPlex Basic Kit (sold separately) is required to run the assay, as Simplex kits include only assay-specific components and do not contain any buffer or plate.

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex assays are bead-based immunoassays designed for multiplex protein analysis on Luminex™ xMAP instruments. They enable the simultaneous detection and quantification of multiple protein targets from a single, small-volume sample, helping researchers save time, sample, and cost compared to traditional single-analyte ELISAs.

Depending on the analyte, ProcartaPlex assays are based on either sandwich ELISA or competitive immunoassay principles.

  • Sandwich assays use two highly specific antibodies binding to different epitopes of the target protein.
  • Competitive assays measure small molecules or analytes with limited epitopes, where the sample analyte competes with a labeled tracer for antibody binding, resulting in an inverse signal.

How the technology works

ProcartaPlex assays use Luminex xMAP technology, in which microscopic magnetic beads are internally dyed with precise ratios of red and infrared fluorophores. Each bead population has a unique spectral signature and is coated with antibodies specific to a single target. This allows different analytes to be measured in parallel within the same well.

Unlike traditional ELISA, where antibodies are immobilized on a plate, ProcartaPlex reagents are free-floating in solution, improving reaction kinetics and enabling efficient multiplexing with the following benefits:

  • Multiplexing efficiency—measure up to 80 protein targets simultaneously
  • Low sample volume—as little as 25 μL for serum or plasma, or 50 μL for cell culture supernatant
  • Time-saving workflow—results in approximately four hours
  • High Throughput - 96-well and even 384-well assay formulations are available helping conserve valuable samples while increasing assay efficiency.

ProcartaPlex multiplex panels and simplex assays are available for six (species: human, mouse, rat, nonhuman primate, porcine, and canine).

For more information and a full list of available targets, visit thermofisher.com/procartaplex.

For Research Use Only.
Specifications
Assay RangeAs determined for Lot 1: 3,250 to 0.8 pg/mL
Assay SensitivityAs determined for Lot 1: 0.644 pg/mL
Bead RegionBeta Endorphin Competitive [55]
Bead TypeMagnetic
For Use With (Equipment)Luminex Instruments
FormatSimplex Kit
GenePOMC
Gene ID (Entrez)5443
Interassay CVAs determined for Lot 1: 7.4%
Intraassay CVAs determined for Lot 1: 7.1%
Product LineProcartaPlex
Sample TypeSerum, Plasma, Cell Culture Supernatants, Cerebrospinal Fluid
Sample VolumeSerum, Plasma, CSF: 25 μL, CCS: 50 μL
Shipping ConditionWet Ice
UniProt IDP01189
CombinabilityCombinable
Product TypeMagnetic Bead-based Assay
Quantity96 Tests
Research AreaEndocrinology
SpeciesHuman
Unit SizeEach