Sensititre™ Equine EQUIN1F Vet AST Plate
Sensititre™ Equine EQUIN1F Vet AST Plate
Sensititre™ Equine EQUIN1F Vet AST Plate
Thermo Scientific™
Sensititre™ Equine EQUIN1F Vet AST Plate
Perform accurate AST with an equine-specific formulary with this convenient plate.
Have Questions?
Catalog NumberQuantity
EQUIN1F10/Pk.
Catalog number EQUIN1F
Price (USD)
-
Quantity:
10/Pk.
Perform accurate AST with an equine-specific formulary with the Thermo Scientific™ Sensititre™ Equine EQUIN1F AST Plate.
Achieve reliable, consistent results for non-fastidious Gram-positive and Gram-negative isolates of veterinary origin with Sensititre Equine EQUIN1F AST Plate.
  • Extended shelf life, room temperature storage eliminates inventory control concerns.
  • Individual packaging allows the laboratory to test one plate at a time with no waste.
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology.
  • Automated or manual read options are available for ultimate flexibility.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
For Veterinary Use Only.
For Veterinary Use Only.
Specifications
Antibiotic CodeVarious
Antimicrobial AgentsVarious
DescriptionSensititre Equine MIC Plate
Isolates Per Plate1
Quantity10/Pk.
Reading OptionsAutoread or manual
Specimen TypeEquine Isolates
Product TypeAST Plate
SpeciesEquine
Target Organism ClassNon-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Test TypeVeterinary AST, Minimum Inhibitory Concentration (MIC)
Unit SizeEach