Does Thermo Fisher Scientific offer an ELISA kit for the detection of IL-8 in rat and mouse?
After much effort, the search for an IL-8 homologue in rats and mice has been unsuccessful. This has led many researchers to conclude that the role of IL-8 in rats and mice is assumed by other chemokines (Broaddus, VA and Hebert, CA, 1996). However, it has been demonstrated that when an anti-human IL-8 antibody was given intratracheally to rats, the measure of lung inflammation and injury were significantly attenuated (Mulligan, MS et al, 1993). In addition, the authors obtained positive immunostaining with the anti-IL-8 antibody on rat pulmonary artery endothelial cells stimulated by TNF-alpha and in the rat lung after immune complex deposition. The authors were unable to determine what cytokine was being neutralized by the antibody. CINC-1 (a rat CXC chemokine and homologue of human MGSA/GRO) was ruled out since the antibody utilized was shown not to cross-react with that chemokine. These data suggest that a rat IL-8-like molecule which is distinct from CINC-1 exists and plays an important role in neutrophil recruitment.
In addition to CINC-1, three other rat CXC chemokines have been identified: CINC-2 alpha, CINC-2 beta, and CINC-3 (GRO beta/MIP-2) (Richmond, A and Shattuck, RL, 1996; Nakagawa, H et al., 1994). To date, the IL-8 homologue has not been identified, but recent data from Lee, J et al. suggest that it may be MIP-2 in rats. The homolog in mice may be KC. We offer an ELISA kit for the detection of rat MIP-2: catalog number ERCXCL2.
References:
1. Broaddus, V.A. and C.A. Hebert (1996) The Role of IL-8 in Inflammatory Diseases, in Chemoattractant Ligands and Their Receptors. Horuk, R., Ed., CRC Press, New York, pp.16-18.
2. Mulligan, M.S., M.L. Jones, M.A. Bolanowski, M.P. Baganoff, C.L. Deppeler, D.M.Meyers, U.S. Ryan, and P.A. Ward (1993) Inhibition of lung inflammatory reactions in rat by antihuman IL-8 antibody. J. Immunol. 150:5585-5595.
3. Richmond, A. and R.L. Shattuck (1996) Melanoma growth stimulatory activity: Physiology, biology, structure/function, and role in disease, in Chemoattractant Ligands and Their Receptors. Horuk, R., Ed., CRC Press, New York, pp. 89-90.
4. Nakagawa, H., N. Komorita, F. Shibata, A. Ikesue, K. Konishi, M. Fujioka, and H. Kato (1994) Identification of cytokine-induced neutrophil chemoattractants (CINC), rat GRO/CINC-2 alpha and CINC-2 beta, produced by granulation tissue in culture: purification, complete amino acid sequences and characterization. Biochem. J. 301:545-550.
5. Lee, J., G. Cacalano, T. Camerato, K. Toy, M.W. Moore, and W.I. Wood (1995) Chemokine binding and activities mediated by the mouse IL-8 receptor. J. Immunol.155:2158-2164.
In addition to CINC-1, three other rat CXC chemokines have been identified: CINC-2 alpha, CINC-2 beta, and CINC-3 (GRO beta/MIP-2) (Richmond, A and Shattuck, RL, 1996; Nakagawa, H et al., 1994). To date, the IL-8 homologue has not been identified, but recent data from Lee, J et al. suggest that it may be MIP-2 in rats. The homolog in mice may be KC. We offer an ELISA kit for the detection of rat MIP-2: catalog number ERCXCL2.
References:
1. Broaddus, V.A. and C.A. Hebert (1996) The Role of IL-8 in Inflammatory Diseases, in Chemoattractant Ligands and Their Receptors. Horuk, R., Ed., CRC Press, New York, pp.16-18.
2. Mulligan, M.S., M.L. Jones, M.A. Bolanowski, M.P. Baganoff, C.L. Deppeler, D.M.Meyers, U.S. Ryan, and P.A. Ward (1993) Inhibition of lung inflammatory reactions in rat by antihuman IL-8 antibody. J. Immunol. 150:5585-5595.
3. Richmond, A. and R.L. Shattuck (1996) Melanoma growth stimulatory activity: Physiology, biology, structure/function, and role in disease, in Chemoattractant Ligands and Their Receptors. Horuk, R., Ed., CRC Press, New York, pp. 89-90.
4. Nakagawa, H., N. Komorita, F. Shibata, A. Ikesue, K. Konishi, M. Fujioka, and H. Kato (1994) Identification of cytokine-induced neutrophil chemoattractants (CINC), rat GRO/CINC-2 alpha and CINC-2 beta, produced by granulation tissue in culture: purification, complete amino acid sequences and characterization. Biochem. J. 301:545-550.
5. Lee, J., G. Cacalano, T. Camerato, K. Toy, M.W. Moore, and W.I. Wood (1995) Chemokine binding and activities mediated by the mouse IL-8 receptor. J. Immunol.155:2158-2164.