Rat L-Selectin (SELL/CD62L) ELISA Kit - FAQs
Rat L-Selectin (SELL/CD62L) ELISA Kit - FAQs
View additional product information for Rat L-Selectin (SELL/CD62L) ELISA Kit - FAQs (ERSELLX10, ERSELL, ERSELLX5)