Rat TIMP-1-Cell Lysates ELISA Kit - FAQs
Rat TIMP-1-Cell Lysates ELISA Kit - FAQs
View additional product information for Rat TIMP-1-Cell Lysates ELISA Kit - FAQs (ERTIMP1CL, ERTIMP1CLX5, ERTIMP1CLX10)