EASY-Spray™ C18 Reversed Phase HPLC Column, 2 μm, 50 μm x 150 mm - FAQs
View additional product information for EASY-Spray™ HPLC Columns - FAQs (ES907, ES905, ES906, ES900, ES911, ES903, ES904, ES901, ES902)
1 product FAQs found
How should I store Easy-Spray HPLC columns when not in use?
When not in use, store Easy-Spray HPLC columns in a cool, dry place. Use the provided end caps to prevent contamination and drying out of the column bed.