ELISA Systems™ Mustard Seed Protein Residue Detection kit for Food Allergen Testing
ELISA Systems™ Mustard Seed Protein Residue Detection kit for Food Allergen Testing
ELISA Systems™ Mustard Seed Protein Residue Detection kit for Food Allergen Testing
ELISA Systems™ Mustard Seed Protein Residue Detection kit for Food Allergen Testing
Thermo Scientific™

ELISA Systems™ Mustard Seed Protein Residue Detection kit for Food Allergen Testing

Rapidly detect and quantify mustard seed protein residue in food and environmental samples.
Have Questions?
카탈로그 번호수량
ESMUS48 Tests
카탈로그 번호 ESMUS
제품 가격(KRW)
-
수량:
48 Tests
Simplify allergen testing with ELISA Systems™ Mustard Seed Protein Residue Detection Kit which helps ensure compliance with food product labeling requirements. The enzyme-linked immunosorbent assay is designed to deliver simple, sensitive, and fast detection and quantification of mustard seed protein material in food and environmental samples. This complete, ready-to-use kit is developed and optimized for rapid and reliable test results that significantly reduce the time required to screen appropriate food products for the presence of mustard seed protein residue.
The European Directive 2003/89/EC includes mustard as a declared food allergen, and as such, it must be included, where applicable, under the appropriate labelling laws regarding food allergens. Mustard is widely used as a flavoring agent, as well as for its texture-enhancing and emulsification capacity in various food preparations, particularly with salad dressings and meats.

Use ELISA Systems Mustard Seed Protein Residue Kit for rapid detection and quantification of mustard seed protein material in food and environmental samples.

  • Fast: Sample preparation to result in under 60 minutes for rapid test turnaround and actions
  • Simple: Ready-to-use liquid reagents and concentrates, plus common handling procedure for all kits, make ELISA Systems Assays easy to implement and run
  • Robust: Ready-to-use liquid controls and allergen-free non-control reagents mean reduced risk of cross contamination that could lead to false-positive results
  • Flexible: Use for qualitative or quantitative testing—a single kit for more applications
  • Sensitive: Detect proteins or protein residues, including heat stable proteins, at low levels for improved assurance of product safety
  • Supported: Our experienced technical team is ready to assist you and help mitigate delays in test reporting


Two common and closely related species of mustard seeds were used in the development of this assay: White Mustard ( Brassica alba, sometimes known as Sinapis alba or Yellow Mustard) and Black Mustard ( Brassica nigra). The assay has been shown to detect the seed proteins from these species, as well as seed protein from another mustard seed: Brassica juncea (Brown Mustard).

All results should be interpreted as part of a HACCP plan for food allergens. Any sample returning a positive result should be regarded as a presumptive result, and confirmation or further testing should be performed.

Not all products are available for sale in all territories. Please inquire.

ELISA Systems™ Food Allergen Test Kits are manufactured by ELISA Systems Pty Ltd.

ELISA Systems is a trademark of ELISA Systems Pty Ltd.
사양
설명ELISA Systems Mustard Seed Protein Residue Detection Kit
검출 가능한 분석물질Mustard Seed Protein Residue
키트 구성품20X wash buffer solution concentrate, Enzyme conjugate, Negative Standard, Positive Standards, Stop solution, Substrate, Test strips
제품 유형Mustard Seed Protein Residue Detection Kit
수량48 Tests
보관 요구 사항2°C to 8°C
이뮤노어세이 키트 형식ELISA (Standard)
Unit SizeEach