Heratherm Cámara Climática 750 L
Heratherm Cámara Climática 750 L
Heratherm Cámara Climática 750 L
Heratherm Cámara Climática 750 L
Heratherm Cámara Climática 750 L
Heratherm Cámara Climática 750 L
Heratherm Cámara Climática 750 L
Heratherm Cámara Climática 750 L
Thermo Scientific™

Heratherm Cámara Climática 750 L

Obtenga confianza en su producto al realizar pruebas de estabilidad y vida útil con las robustas Cámaras climáticas Heratherm diseñadas para cumplir con una variedad de demandas de control de calidad.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoModelo de puertaCapacidad (métrico)
ESS750Sólido (exterior), vidrio (interior)750 L
ESG750Vidrio750 L
ESS250Solid outer door with inner glass door247 L
ESG250Glass door247 L
Número de catálogo ESS750
Precio (MXN)
-
Modelo de puerta:
Sólido (exterior), vidrio (interior)
Capacidad (métrico):
750 L
Temperature and humidity controls, well suited for stability testing, shelf life testing, and environmental testing.
  • Broad relative humidity range to cover many applications, including humidity levels below ambient.
  • Environmentally friendly water-efficient humidification system.
  • Minimal condensate in standard applications.
  • Integrated water tank; fixed water connection for in-house purified water option available.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
Especificaciones
Tipos de alarmaAudible and Visible
Capacidad (imperial)26.5 cu. ft.
Capacidad del estante (imperial)66.1 lbs
Capacidad del estante (métrico)30 kg
Certificaciones/conformidadUL, CE
Salidas de datosUSB, Ethernet
Fondo (imperial)34.84 in.
Fondo interior (imperial)24.8 in.
Fondo (métrico)885 mm
Fondo interior (métrico)630 mm
Pantalla7 in. touchscreen user interface
Modelo de puertaSólido (exterior), vidrio (interior)
Requisitos eléctricos100/240V, 50/60 Hz, 1400W
Altura (imperial)72.24 in.
Altura (imperial) interior57.09 in
Altura (métrico)1835 mm
Altura (métrico) interior1450 mm
Rango de humedad10 - 90% RH
Resolución de humedad0.1% RHNo
N.º de ruedas giratorias4
Tipo de tapónNema 5-20P
Puertos2 access ports
ProgramableTemperature and humidity ramping
Área del estante (imperial) 4.9 ft²
Área del estante (métrico)0.46 m²
Material del estanteStainless Steel (304/1.4301)
Peso con embalaje (imperial)840 lbs
Peso con embalaje (métrico)381 kg
Control de temperaturaMicroprocessor - programmable
Resolución de temperatura0,1 °C
Estabilidad de la temperatura≤ ±0.1°C @ 20°C to 37°C
Uniformidad de temperatura≤ ±0.5°C @ 20°C to 37°C
TipoCámara de humedad
Garantía24 months
Consumo de agua0.15 to 1 L per day
Resistividad del agua0.05 - 1 MΩ
Vataje1400 W
Anchura (imperial)42.52 in.
Anchura (imperial) interior33.07 in.
Anchura (imperial) del estante22.6 in.
Anchura (métrico)1080 mm
Anchura (métrico) interior840 mm
Anchura (métrico) del estante795 mm
Capacidad (métrico)750 L
Material de la cámaraPowder-coated Cold Rolled Steel
MaterialPowder-coated Cold Rolled Steel
N.º de estantes3
Humedad relativa10 to 90%
Intervalo de temperatura (métrico)0 to 70°C
Tensión100/115/220/230 V
Unit SizeEach