Revco™ ExF -86°C Upright Ultra-Low Temperature Freezer, 28 cu. ft. (792.8L) capacity, holds 600 boxes, 230V/50Hz, 12A, painted galvaneel interior - FAQs
Revco™ ExF -86°C Upright Ultra-Low Temperature Freezer, 28 cu. ft. (792.8L) capacity, holds 600 boxes, 230V/50Hz, 12A, painted galvaneel interior - FAQs
View additional product information for Revco™ ExF -86°C Upright Ultra-Low Temperature Freezers - FAQs (EXF60086V)