PEEK Fingertight Fittings
Actual product may vary
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
Thermo Scientific™
PEEK Fingertight Fittings
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDescription
F-300XPEEK Wing Nut w/PEEK Ferrule1/16 in.
U-467RXColumn End Plugs, Red1/16 in., 10-32 Coned, Delrin, Red
U-467BLKShort Head Port Plug, Black1/16 in., 10-32 Coned, Delrin, Black
Catalog number F-300X
Price (USD)
141.23
Pack of 10
Type:
PEEK Wing Nut w/PEEK Ferrule
Description:
1/16 in.
Price (USD)
141.23
Pack of 10
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
Description
  • Resist cracking, breaking, thread stripping and leaking in both low and high pressure applications
  • Biocompatible for a broad range of applications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description1/16 in.
Quantity10 Pack
For Use With (Equipment)LC Tubing
MaterialPEEK
TypePEEK Wing Nut w/PEEK Ferrule
Unit SizePack of 10