HyPerforma™ Rocker Bioreactor, with load cells
Thermo Scientific™
HyPerforma™ Rocker Bioreactor, with load cells
The Thermo Scientific™ HyPerforma Rocker Bioreactor brings control and measurement to rocking bioreactor applications.
Have Questions?
Catalog NumberMaterial
F100-2683-001Stainless Steel
Catalog number F100-2683-001
Price (USD)
-
Material:
Stainless Steel
The Thermo Scientific™ HyPerforma Rocker Bioreactor brings control and measurement to rocking bioreactor applications. The rocker is controlled by a HyPerforma G3Lab Controller and TruBio software, providing a complete solution for research, process development, or seed train production applications. The rocker is compatible with a 10 L, 20 L or 50 L Thermo Scientific™ Rock-it BioProcess Container (BPC), which provides from 5 L to 25 L of working volume and integrates a novel TruFluor pH+dO2 sensor for combined pH, DO , and temperature monitoring.
HyPerforma Rocker Bioreactor features include:
  • Rocking motion is configurable—from a smooth waveform that minimizes shear forces for sensitive cell lines, through four intermediate steps, to an aggressive motion that maximizes oxygen transfer for robust cells with high oxygen demands
  • Optional heating blanket—includes bimetallic temperature limiting switch embedded in the blanket to help protect against overheating or fires
  • Quick, simple setup with G3Lab Controller and TruBio software
  • TruFluor pH+dO2 sensor provides measurement and control of critical process parameters: pH, DO, and temperature
Applications
  • Suspension and adherent cell culture processes
  • Fermentation processes
  • PD scale to full cGMP manufacturing workflows
Specifications
Dimensions(Hx W x D) 264 x 782 x 701 mm (10.4 x 30.8 x 27.6 in), 490 mm x 835 mm x 712 mm (490.3 x 32.9 x 28.0 in.) with cover
Max. Voltage110 to 120 V, 220 to 240 V, 50/60 Hz, powered by the G3Lab Controller
Product LineHyPerforma
Temperature Accuracy±0.15°C at 15°C to 40°C (±0.25°F at 59°F to 104°F)
Weight38.5 kg (85 lbs)
MaterialStainless Steel
Unit SizeEach
Contents & Storage
• HyPerforma Rocker Bioeactor
• Vent filter assembly
• Cable assembly
• Rocker cover, base, and tray
• Heat only, with load cells