Phire Green Hot Start II DNA Polymerase, 1,000 reactions - FAQs
View additional product information for Phire Hot Start II DNA Polymerase - FAQs (F122L, F124S, F122S, F124L)
3 product FAQs found
Do Phire and Phusion Hot Start II DNA Polymerases need a separate activation step in the PCR protocol?
No separate activation step is required since Phire and Phusion Hot Start II DNA Polymerases are inactivated by a reversibly bound, specific Affibody ligand that dissociates during initial denaturation.
Can I use Phusion Green and Phire Green PCR products for subsequent applications such as sequencing, ligation, and restriction digestion?
Yes. The dyes do not interfere with downstream applications such as DNA sequencing, ligation, and restriction digestion.
What dyes are present in Phusion Green and Phire Green buffers?
Phusion Green and Phire Green buffers contain two dyes for monitoring electrophoresis progress. The blue dye migrates with 3-5 kb DNA fragments and the yellow dye migrates faster than 10 bp DNA fragments in 1% agarose gel. The dyes have excitation peaks at 424 nm and 615 nm, respectively.