Phire Hot Start II DNA Polymerase
Thermo Scientific™

Phire Hot Start II DNA Polymerase

Thermo Scientific Phire Hot Start II DNA Polymerase is an enhanced PCR enzyme for routine and high throughput PCR applications. It is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of Reactions
F124SGreen200 Reactions
F122SColorless200 Reactions
F122LColorless1000 Reactions
F124LGreen1000 Reactions
Catalog number F124S
Price (USD)
202.65
Online exclusive
241.00
Save 38.35
Each
Color:
Green
No. of Reactions:
200 Reactions
Request bulk or custom format
Price (USD)
202.65
Online exclusive
241.00
Save 38.35
Each
Thermo Scientific Phire Hot Start II DNA Polymerase is an enhanced PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain which allows short extension times (10–15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase (Cat. No. EP0072).

The hot start modification of Phire Hot Start II DNA Polymerase is based on the Affibody inactivation technology. This technology increases the specificity of the PCR reaction with no additional time required for initial activation of the enzyme.

Phire Green format is a combination of Phire Hot Start II DNA Polymerase and 5X Phire Green Buffer. The buffer includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Features

  • No reactivation step with hot start
  • Amplification four times faster than hot start Taq
  • Minimal reaction optimization due to high inhibitor tolerance
  • Abundant products due to high efficiency
  • Amplifies significantly longer DNA fragments than hot start Taq
  • Direct loading on gel with Green Buffer

Applications

  • Hot Start PCR
  • Standard PCR
  • Non-high-fidelity PCR
  • Fast PCR
  • High throughput PCR
  • Genotyping

Notes

  • The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Fidelity (vs. Taq)2X
Hot StartBuilt-In Hot Start
No. of Reactions200 Reactions
OverhangBlunt
PolymerasePhire Hot Start II DNA Polymerase
Product TypeHot Start High-Fidelity DNA Polymerase
Quantity200 reactions
Reaction FormatSeparate Components
Shipping ConditionWet or Dry Ice
Starting MaterialDNA
Volume50 μL
For Use With (Application)Cloning, Hot-start PCR
GC-Rich PCR PerformanceMedium
Reaction SpeedFast
Unit SizeEach
Contents & Storage
• Phire Hot Start II DNA Polymerase, 1 x 200 μL
• 5X Phire Green Reaction Buffer, 2 x 1.5 mL
• DMSO, 1 x 500 μL

Store at -20°C.

Frequently asked questions (FAQs)

Do Phire and Phusion Hot Start II DNA Polymerases need a separate activation step in the PCR protocol?

No separate activation step is required since Phire and Phusion Hot Start II DNA Polymerases are inactivated by a reversibly bound, specific Affibody ligand that dissociates during initial denaturation.

Can I use Phusion Green and Phire Green PCR products for subsequent applications such as sequencing, ligation, and restriction digestion?

Yes. The dyes do not interfere with downstream applications such as DNA sequencing, ligation, and restriction digestion.

What dyes are present in Phusion Green and Phire Green buffers?

Phusion Green and Phire Green buffers contain two dyes for monitoring electrophoresis progress. The blue dye migrates with 3-5 kb DNA fragments and the yellow dye migrates faster than 10 bp DNA fragments in 1% agarose gel. The dyes have excitation peaks at 424 nm and 615 nm, respectively.