Phire Hot Start II PCR Master Mixes
Thermo Scientific™

Phire Hot Start II PCR Master Mixes

Thermo Scientific Phire Hot Start II PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phire Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of Reactions
F126SGreen200 Reactions
F125SColorless200 Reactions
F125LColorless1000 Reactions
F126LGreen1000 Reactions
Catalog number F126S
Price (USD)
294.65
Online exclusive
335.00
Save 40.35
Each
Color:
Green
No. of Reactions:
200 Reactions
Request bulk or custom format
Price (USD)
294.65
Online exclusive
335.00
Save 40.35
Each
Thermo Scientific Phire Hot Start II PCR Master Mix is a convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phire Hot Start II DNA Polymerase, nucleotides, and optimized reaction buffer, including MgCl2. Only template and primers need to be added to the PCR reaction.

Phire Green Hot Start II PCR Master Mix also includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Phire Hot Start II DNA Polymerase, included in the master mix, is an enhanced PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain that allows short extension times (10–15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase (Cat. No. EP0072).

Features

  • No reactivation step with quick hot start
  • Amplify four times faster than with hot start Taq
  • Minimal reaction optimization due to high inhibitor tolerance
  • Abundant products due to high efficiency
  • Amplify significantly longer DNA fragments than with any hot start Taq
  • Direct loading on gel with Green format

Applications

  • Hot Start PCR
  • Routine PCR
  • Non-high-fidelity PCR
  • Fast PCR
  • High throughput PCR
  • Genotyping

Notes

  • The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Fidelity (vs. Taq)2X
Hot StartBuilt-In Hot Start
No. of Reactions200 Reactions
OverhangBlunt
PolymerasePhire Hot Start II DNA Polymerase
Product TypeHot Start PCR Master Mix
Quantity200 reactions
Reaction FormatSuperMix or Master Mix
Shipping ConditionWet or Dry Ice
Starting MaterialDNA
Concentration2X
For Use With (Application)Cloning, Hot-start PCR
GC-Rich PCR PerformanceMedium
Reaction SpeedFast
Unit SizeEach
Contents & Storage
• 2X Phire Green Hot Start II PCR Master Mix, 4 x 1.25 mL
• DMSO, 1 x 0.5 mL
• Nuclease-free water, 4 x 1.25 mL

Store at -20°C.

Frequently asked questions (FAQs)

Do Phire and Phusion Hot Start II DNA Polymerases need a separate activation step in the PCR protocol?

No separate activation step is required since Phire and Phusion Hot Start II DNA Polymerases are inactivated by a reversibly bound, specific Affibody ligand that dissociates during initial denaturation.

Can I use Phusion Green and Phire Green PCR products for subsequent applications such as sequencing, ligation, and restriction digestion?

Yes. The dyes do not interfere with downstream applications such as DNA sequencing, ligation, and restriction digestion.

What dyes are present in Phusion Green and Phire Green buffers?

Phusion Green and Phire Green buffers contain two dyes for monitoring electrophoresis progress. The blue dye migrates with 3-5 kb DNA fragments and the yellow dye migrates faster than 10 bp DNA fragments in 1% agarose gel. The dyes have excitation peaks at 424 nm and 615 nm, respectively.