Fluo-4 AM - Packaged For High-Throughput Screening
Invitrogen™

Fluo-4 AM - Packaged For High-Throughput Screening

Fluo-4 AM - packaged for high-throughput screening.

Have Questions?
Catalog NumberQuantity
F142025 x 1 mg
Catalog number F14202
Price (USD)
1,788.00
Each
Quantity:
5 x 1 mg
Price (USD)
1,788.00
Each

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. Fluo-3 has been used to image the spatial dynamics of Ca2+ signaling, in flow cytometry experiments involving photoactivation of caged chelators, second messengers, and neurotransmitters, and for cell-based pharmacological screening. Fluo-4 is an analog of fluo-3 with the two chlorine substituents replaced by fluorines, which results in increased fluorescence excitation at 488 nm and consequently higher fluorescence signal levels. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. These indicators are useful for fluorescence and confocal microscopy, flow cytometry, and microplate screening applications.

Calcium Indicator (AM Ester) Specifications:

  • Label (Ex/Em of Ca2+–bound form): Fluo-4 (494/506 nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in buffer: ∼335 nM
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

Using TPEN to Control Heavy Metal Cations

In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator TPEN (Cat. No. T1210).

More Choices for Fluorescent Calcium Indicators

We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeFluo-4
Emission Wavelength516 nm
Excitation Wavelength494 nm
Quantity5 x 1 mg
Shipping ConditionRoom Temperature
For Use With (Application)Calcium Indicator
For Use With (Equipment)Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
Product TypeDye
Unit SizeEach
Contents & Storage
Store at -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

Can I fix my cells after loading with Fluo-4 AM dye for the detection of calcium?

No. Since Fluo-4 AM isn't covalently bound to any cellular components and fixation compromises the membrane, the dye would not be retained by the cell.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am doing calcium flux imaging with your Fura-2 calibration kit, but am seeing a large variability in ratio in different places around the slide. I am correcting for uniform illumination, using the product as directed, and sealing the coverslip with nail polish.

The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I need to label cells with Fluo-4, AM, for a calcium flux assay. How long after labeling will the dye be retained?

After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (317)

Citations & References
Abstract
Functional implications of calcium permeability of the channel formed by pannexin 1.
Authors:Vanden Abeele F,Bidaux G,Gordienko D,Beck B,Panchin YV,Baranova AV,Ivanov DV,Skryma R,Prevarskaya N
Journal:The Journal of cell biology
PubMed ID:16908669
Although human pannexins (PanX) are homologous to gap junction molecules, their physiological function in vertebrates remains poorly understood. Our results demonstrate that overexpression of PanX1 results in the formation of Ca(2+)-permeable gap junction channels between adjacent cells, thus, allowing direct intercellular Ca(2+) diffusion and facilitating intercellular Ca(2+) wave propagation. More ... More
Impaired insulin secretion and glucose intolerance in synaptotagmin-7 null mutant mice.
Authors:Gustavsson N,Lao Y,Maximov A,Chuang JC,Kostromina E,Repa JJ,Li C,Radda GK,Südhof TC,Han W
Journal:Proceedings of the National Academy of Sciences of the United States of America
PubMed ID:18308938
Vertebrates express at least 15 different synaptotagmins with the same domain structure but diverse localizations and tissue distributions. Synaptotagmin-1,-2, and -9 act as calcium sensors for the fast phrase of neurotransmitter release, and synaptotagmin-12 acts as a calcium-independent modulator of release. The exact functions of the remaining 11 synaptotagmins, however, ... More
High-throughput microfluidic mixing and multiparametric cell sorting for bioactive compound screening.
Authors:Young SM, Curry MS, Ransom JT, Ballesteros JA, Prossnitz ER, Sklar LA, Edwards BS
Journal:J Biomol Screen
PubMed ID:15006133
HyperCyt, an automated sample handling system for flow cytometry that uses air bubbles to separate samples sequentially introduced from multiwell plates by an autosampler. In a previously documented HyperCyt configuration, air bubble separated compounds in one sample line and a continuous stream of cells in another are mixed in-line for ... More
Drosophila Hsc70-4 is critical for neurotransmitter exocytosis in vivo.
Authors:Bronk P, Wenniger JJ, Dawson-Scully K, Guo X, Hong S, Atwood HL, Zinsmaier KE
Journal:Neuron
PubMed ID:11395008
'Previous in vitro studies of cysteine-string protein (CSP) imply a potential role for the clathrin-uncoating ATPase Hsc70 in exocytosis. We show that hypomorphic mutations in Drosophila Hsc70-4 (Hsc4) impair nerve-evoked neurotransmitter release, but not synaptic vesicle recycling in vivo. The loss of release can be restored by increasing external or ... More
Multiplex GPCR assay in reverse transfection cell microarrays.
Authors:Mishina YM, Wilson CJ, Bruett L, Smith JJ, Stoop-Myer C, Jong S, Amaral LP, Pedersen R, Lyman SK, Myer VE, Kreider BL, Thompson CM
Journal:J Biomol Screen
PubMed ID:15140381
'G protein-coupled receptors (GPCRs) are a superfamily of proteins that include some of the most important drug targets in the pharmaceutical industry. Despite the success of this group of drugs, there remains a need to identify GPCR-targeted drugs with greater selectivity, to develop screening assays for validated targets, and to ... More