Phire Plant Direct PCR Master Mix
Thermo Scientific™

Phire Plant Direct PCR Master Mix

Thermo Scientific Phire Plant Direct PCR Master Mix is designed to amplify DNA directly from plant samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
F160S250 Reactions
F160L1250 Reactions
Catalog number F160S
Price (USD)
177.65
Online exclusive
207.00
Save 29.35
Each
No. of Reactions:
250 Reactions
Request bulk or custom format
Price (USD)
177.65
Online exclusive
207.00
Save 29.35
Each
Thermo Scientific Phire Plant Direct PCR Master Mix is designed to amplify DNA directly from plant samples. No DNA purification is required prior to PCR. The kit is based on Phire Hot Start II DNA Polymerase, a specially engineered enzyme with a DNA binding domain. The unique features of this DNA polymerase make it extremely robust and tolerant of many PCR inhibitors present in plant material like complex polysaccharides, polyphenols, and others.

Phire Plant Direct PCR Master Mix has been tested with leaves and seeds from a wide variety of plant species as well as fungi, bacteria, and yeast. The kit includes a complete set of optimized reagents and detailed protocols, making it an ideal choice for amplification of plant DNA.

Features

  • Direct PCR allows sample to be added directly to PCR reaction, reducing time-consuming and expensive DNA purification steps
  • Available Dilution & Storage protocol allows several PCR reactions from one sample
  • Specially engineered Phire Hot Start II DNA Polymerase for extremely short PCR protocol times
  • Master mix format with premixed gel loading dye minimizes risk of cross-contamination, reduces sample handling, and allows direct loading on gel

Applications

  • Genotyping
  • Transgene detection
  • Knockout analysis
  • Sequencing

Notes

  • Annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Hot StartBuilt-In Hot Start
Includes Label or DyeYes
No. of Reactions250 Reactions
OverhangBlunt
PolymerasePhire Hot Start II DNA Polymerase
Product TypeDirect PCR Master Mix
Quantity250 reactions
Reaction FormatSuperMix or Master Mix
Sample TypePlant
Shipping ConditionWet or Dry Ice
Starting MaterialDNA
Concentration2X
For Use With (Application)Genotyping, Hot-start PCR
GC-Rich PCR PerformanceMedium
PCR MethodDirect PCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
• 2X Phire Plant Direct PCR Master Mix, 2 x 1.25 mL
• Dilution Buffer, 1 x 5 mL
• 25 μM Control Primer Mix, 40 μL
• Nuclease-free water, 2 x 1.25 mL
• O'GeneRuler Express DNA Ladder, 50 μg (sufficient for 100 applications)

Store at -20°C.

Frequently asked questions (FAQs)

Can I use frozen leaves with the Phire Plant Direct PCR Master Mix or Kit?

Yes, frozen (-20 degrees C and -70 degrees C), dried and fresh plant leaves can be used. We have succeeded in amplifying DNA directly from oak leaves stored at -20 degrees C for 10 years. However the highest amplification yields are achieved when using fresh samples.