When would I use the FluxOR Red Potassium Ion Channel Assay in lieu of the FluxOR II Green Potassium Ion Channel Assay?
The FluxOR Red Potassium Ion Channel Assay is recommended for cells that are expressing GFP or related fluorescent proteins or with cells that have high autofluorescence in the green emission channel.