FluoSpheres™ Polystyrene Microspheres, 15 μm, green fluorescent (450/480), for blood flow determination
Invitrogen™

FluoSpheres™ Polystyrene Microspheres, 15 μm, green fluorescent (450/480), for blood flow determination

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed fromRead more
Have Questions?
Catalog NumberQuantity
F21010
also known as F-21010
10 mL
Catalog number F21010
also known as F-21010
Price (USD)
564.00
Each
Quantity:
10 mL
Price (USD)
564.00
Each
Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Our Molecular Probes™ FluoSpheres™ beads are manufactured using high-quality, ultraclean polystyrene and are loaded with a variety of our proprietary dyes to create intensely fluorescent beads that typically show little or no photobleaching, even when excited with the intense illumination required for fluorescence microscopy.

FluoSpheres™ microspheres in the 10–15 μm size range were designed specifically for regional blood flow studies in tissues and organs. Typically for these types of studies, the microspheres are injected at desired locations in the circulatory system and eventually lodge in the capillaries, where they can be extracted and the fluorescence quantified. The use of fluorescent microspheres eliminates the radiation hazards and disposal hurdles associated with radiolabeled microspheres. Multiple (up to seven or more with good spectral correction) simultaneously circulating FluoSpheres™ microspheres can be discriminated. Microspheres that have been perfused or injected into tissues and organs retain their fluorescence following histological serial sectioning. Our FluoSpheres™ microspheres for blood flow determination, which are available in eleven distinguishable fluorescent colors, are also compatible with blood flow analyzer systems that perform automated extraction and analysis.

FluoSpheres™ Microsphere Specifications:

• Label (Ex/Em): Green fluorescent (450/480)
• Nominal bead diameter: 15 μm

Choices for FluoSpheres™ Fluorescent Microspheres
In addition to microspheres for blood flow studies, investigate our complete offering of fluorescent microspheres products. Among those products you’ll find beads with these variations:
• Ten fluorescent colors
• Ten nominal bead diameters: 0.02 μm, 0.04 μm, 0.1 μm, 0.2 μm, 0.5 μm, 1.0 μm, 2.0 μm, 4.0 μm, 10.0 μm, and 15.0 μm
• Four surface modifications for protein coupling: carboxylate, sulfate, aldehyde-sulfate, amine
• Microspheres that are additionally precoupled with streptavidin, NeutrAvidin, biotin, europium, and platinum

Choices for Unstained Microspheres
We also offer hundreds of choices for UltraClean™ surfactant-free microspheres for research and commercial applications.

We’ll Make a Custom Microsphere Product for You
We will prepare custom orders upon request. For example, FluoSpheres™ beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1.0 x 106 beads/mL
Product LineFLUOSPHERES
Quantity10 mL
Shipping ConditionRoom Temperature
ColorGreen
Diameter (Metric)15 μm
MaterialPolystyrene
Product TypeMicrosphere
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

I want to use fluorescent microspheres for blood tracing studies. What functional group should be on the surface of the beads?

We offer FluoSpheres fluorescent microspheres that are available in 10 and 15 µm diameters, with various wavelengths, for tracer studies. If those sizes or available colors are not optimal for your system, then we recommend the carboxylate-modified microspheres, which are available in a wide range of sizes and colors.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the warranty for FluoSpheres microspheres?

The warranty period for FluoSpheres microspheres is 1-year from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

After washing and centrifugation, there was only a very small pellet left of my microsphere beads and the solution was transparent. Why is this?

Centrifugation is not an effective way to collect smaller microspheres; many particles remain in the solution even if you can visualize a small pellet. For beads less than 1 µm in diameter, we recommend washing by either:

Cross-flow filtration, as these particles have a very high compression modulus and can withstand high g-forces without risk of harm or dialysis with a 500 kDa MWCO
Note: Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I've had my microspheres for over a year, and I'm wondering if they're still good to use. What are some good ways to check their functionality?

Bacterial contamination is the most common cause of microspheres becoming unusable. Many of our particles are supplied with a low level of sodium azide to prevent bacterial contamination, but sometimes this can still occur. Bacterial contamination is best assessed by plating on appropriate growth medium and checking the plates after 72 hr.

Find additional tips, troubleshooting help, and resources within ourMicrospheres Support Center.

I accidentally froze my microspheres; can I still use them?

Even brief freezing can cause irreversible aggregation and potential distortion of the bead shape. You should not use these microspheres.

Find additional tips, troubleshooting help, and resources within our Microspheres Support Center.

Citations & References (5)

Citations & References
Abstract
Use of coherent control methods through scattering biological tissue to achieve functional imaging.
Authors:Dela Cruz JM, Pastirk I, Comstock M, Lozovoy VV, Dantus M
Journal:Proc Natl Acad Sci U S A
PubMed ID:15569924
'We test whether coherent control methods based on ultrashort-pulse phase shaping can be applied when the laser light propagates through biological tissue. Our results demonstrate experimentally that the spectral-phase properties of shaped laser pulses optimized to achieve selective two-photon excitation survive as the laser pulses propagate through tissue. This observation ... More
Detection technologies in proteome analysis.
Authors:Patton WF
Journal:J Chromatogr B Analyt Technol Biomed Life Sci
PubMed ID:12015990
'Common strategies employed for general protein detection include organic dye, silver stain, radiolabeling, reverse stain, fluorescent stain, chemiluminescent stain and mass spectrometry-based approaches. Fluorescence-based protein detection methods have recently surpassed conventional technologies such as colloidal Coomassie blue and silver staining in terms of quantitative accuracy, detection sensitivity, and compatibility with ... More
Visualizing regional myocardial blood flow in the mouse.
Authors:Krueger MA, Huke SS, Glenny RW,
Journal:Circ Res
PubMed ID:23513055
The spatial distribution of blood flow in the hearts of genetically modified mice is a phenotype of interest because derangements in blood flow may precede detectable changes in organ function. However, quantifying the regional distribution of blood flow within organs of mice is challenging because of the small organ volume ... More
Effects of gestational age and cortisol treatment on ovine fetal heart function in a novel biventricular Langendorff preparation.
Authors:Fletcher AJ, Forhead AJ, Fowden AL, Ford WR, Nathanielsz PW, Giussani DA,
Journal:J Physiol
PubMed ID:15513943
Structural and functional maturation of a number of fetal organs and physiological systems occurs in the immediate period prior to term, in association with the prepartum increase in fetal plasma cortisol concentration. At present, little is known about how myocardial sensitivity to adrenergic and muscarinic cholinergic stimulation changes as the ... More
Multiplexed spectral signature detection for microfluidic color-coded bioparticle flow.
Authors:Huang NT, Truxal SC, Tung YC, Hsiao AY, Luker GD, Takayama S, Kurabayashi K,
Journal:Anal Chem
PubMed ID:20979407
Here, we report a high-speed photospectral detection technique capable of discriminating subtle variations of spectral signature among fluorescently labeled cells and microspheres flowing in a microfluidic channel. The key component used in our study is a strain-tunable nanoimprinted grating microdevice coupled with a photomultiplier tube (PMT). The microdevice permits acquisition ... More