750μL Nonsterile Micro-Centrifugal Filters
Thermo Scientific™
750μL Nonsterile Micro-Centrifugal Filters
Filter volumes as low as 50μL with low hold-up volume with these 750μL polypropylene micro-centrifugal filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial (Membrane)Pore Size
F2517-7Regenerated Cellulose0.2 μm
F2517-1Cellulose Acetate0.22 μm
F2517-10PTFE0.45 μm
F2517-2Cellulose Acetate0.45 μm
F2517-8Regenerated Cellulose0.45 μm
Catalog number F2517-7
Price (MXN)
-
Material (Membrane):
Regenerated Cellulose
Pore Size:
0.2 μm
Filter volumes as low as 50μL with low hold-up volume with these Thermo Scientific™ National 750μL Nonsterile Micro-Centrifugal Filters. The filters feature a virgin polypropylene filter housing with tapered 2mL, capped receiver tube, and can be used with any benchtop microcentrifuge.
  • 10,000xG maximum centrifugal force
  • Nonsterile
  • Bulk packed
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionRegenerated Cellulose Membrane; 0.2µm Pore Size
For Use With (Equipment)Centrifuge
Housing MaterialPolypropylene
Product LineTarget2
TypeMicrocentrifugal Filter
Volume (Metric)750 μL
Material (Membrane)Regenerated Cellulose
Pore Size0.2 μm
SterilityNon-sterile
Unit SizeEach