FlexMAP™ 3D™ Performance Verification Kit
Invitrogen™
FlexMAP™ 3D™ Performance Verification Kit
The convenient verification kit contains xMAP® verification reagents used to verify calibration of the MagPlex and MicroPlex classification channels andRead more
Catalog NumberQuantity
F3DPVERK2525 reactions
Catalog number F3DPVERK25
Price (JPY)
114,700
Each
Quantity:
25 reactions
The convenient verification kit contains xMAP® verification reagents used to verify calibration of the MagPlex and MicroPlex classification channels and reporter channel. New Fluidic Controls included for early detection. Lot specific CD is included for importing target values. The kit is intended to be used with the off plate reagent area. Stripwells are included with each kit. Each kit will provide 25 verifications
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineLuminex
Quantity25 reactions
Shipping ConditionWet Ice
For Use With (Equipment)FlexMAP 3D System
Product TypeCalibration Kit
Unit SizeEach
Contents & Storage
Contains 28 strip wells, FM3D Performance Verification Kit CD, Luminex Classification Control Microspheres, MagPlex Classification Control Microspheres, Luminex Reporter Control Microspheres, xMAP Fluidics 1 Microspheres, xMAP Fluidics 2 Microspheres. Store at 2-8°C.
Frequently asked questions (FAQs)
How long does it take to acquire ProcartaPlex assay data on the different Luminex instruments?
With everything working smoothly, here is the approximate read time for one plate on different Luminex instruments:
- FLEXMAP 3D Systems and INTELLIFLEX Systems will read 96 well plates in ~20 min, and 384 well plates in ~75 min.
- Luminex 100/200 Systems will read 96 well plates in ~40 min.
- MAGPIX System will read 96 well plates in ~50-60 min. Please note that these are estimates for the read times of the plates only, and do not include time for calibration or set-up of the instrument.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.