DyNAmo Flash Probe qPCR Kit - FAQs
View additional product information for DyNAmo Flash Probe qPCR Kit - FAQs (F455S)
1 product FAQs found
Do I need to use Fast plastics to run a Fast qPCR reaction?
Specialized Fast PCR plates are not required if you are using a standard qPCR machine with a fast protocol. However, Fast Block PCR Plates (AB-1900) can be purchased which are compatible with the thermal blocks on fast qPCR thermal cyclers.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.