Search
View additional product information for Phusion™ Plus PCR Master Mixes - FAQs (F631S, F631XL, F632S, F632XL, F631L, F632L)
1 product FAQs found
Reaction setup with Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix can be done either on ice or at room temperature.