Phusion™ Plus PCR Master Mix, 100 x 50 μL reactions - FAQs
View additional product information for Phusion™ Plus PCR Master Mixes - FAQs (F631S, F631XL, F632S, F632XL, F631L, F632L)
1 product FAQs found
Can I set up my reactions on ice if I am using the Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix (Cat. No. F631S)?
Reaction setup with Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix can be done either on ice or at room temperature.