Phusion™ Plus PCR Master Mixes
Thermo Scientific™

Phusion™ Plus PCR Master Mixes

Thermo Scientific Phusion Plus PCR Master Mix is a convenient, ready-to-use 2X mixture of Phusion Plus DNA Polymerase, buffer, magnesium chloride, and dNTPs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of Reactions
F631XLColorless2000 Reactions
F632LGreen500 Reactions
F632SGreen100 Reactions
F632XLGreen2000 Reactions
F631LColorless500 Reactions
F631SColorless100 Reactions
Catalog number F631XL
Price (MXN)
-
Color:
Colorless
No. of Reactions:
2000 Reactions
Request bulk or custom format
Thermo Scientific Phusion Plus Green PCR Master Mix is a convenient, ready-to-use 2X mixture of Phusion Plus DNA Polymerase, buffer, magnesium chloride, and dNTPs. The master mix format simplifies reaction setup with minimal pipetting steps and retains all features of Phusion Plus DNA Polymerase. Additionally, two tracking dyes and a density reagent are pre-added to the master mix for direct loading of PCR products on gels.

Features of Phusion™ Plus PCR Master Mix include:

  • High fidelity—provides >100X higher sequence accuracy than Taq polymerase
  • Convenient setup—enables a universal annealing temperature of 60°C in PCR
  • Enhanced specificity—hot-start mechanism reduces non-specific amplification and primer degradation
  • Benchtop stability—automation enabled due to 24-hour stability of assembled reactions
  • High inhibitor tolerance—works with DNA of suboptimal purity

Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing. It is a Pyrococcus-like proofreading DNA polymerase fused to a processivity-enhancing domain, enabling Phusion Plus PCR Master Mix to generate PCR sequences with high accuracy, sensitivity, and inhibitor tolerance. Its novel reaction buffer includes proprietary additives that stabilize primer/template duplexes during the annealing step at 60°C. This unique buffer formulation increases PCR specificity without the need to optimize annealing temperature for each primer pair, helping save time and avoid mistakes in PCR runs. Furthermore, with Phusion Plus PCR Master Mix and its universal annealing feature, different PCR reactions can be cycled together using the same protocol.

Phusion Plus PCR Master Mix is inactive at ambient temperatures due to its hot-start mechanism mediated by Affibody molecules, allowing setup and storage of assembled PCR reactions at room temperature. Enzyme activity is fully restored after the initial denaturation step.

Applications:

  • High-fidelity PCR
  • Molecular cloning and subcloning
  • Template generation for sequencing
  • GC-rich PCR
  • Long-range PCR
  • Multiplex target amplification
  • Synthetic biology and DNA assembly workflows
  • DNA template generation for in vitro transcription and mRNA synthesis
  • Site-directed mutagenesis
  • Microbial genomics and microbiome research
  • Synthetic biology and DNA assembly workflows
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorColorless
Fidelity (vs. Taq)>100X
Hot StartBuilt-In Hot Start
No. of Reactions2000 Reactions
OverhangBlunt
PolymerasePhusion Plus DNA Polymerase
Quantity2000 x 50 μL reactions
Reaction FormatReady to use master mix
Shipping ConditionDry Ice
Size (Final Product)20 kb or less
Concentration2X
For Use With (Application)Hot-start PCR, High-fidelity PCR
GC-Rich PCR PerformanceHigh
Reaction Speed15 to 30 sec./kb
Unit SizeEach
Contents & Storage
Four packs, each containing:
• 2X Phusion Plus PCR Master Mix (10 x 1.25 mL)
• 5X Phusion GC Enhancer (4 x 1.25 mL)
• Nuclease-free water (10 x 1.25 mL)

Store at –15°C to –25°C

Frequently asked questions (FAQs)

Can I set up my reactions on ice if I am using the Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix (Cat. No. F631S)?

Reaction setup with Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix can be done either on ice or at room temperature.