Canine Genotypes Panel 2.1 Kit
Thermo Scientific™

Canine Genotypes Panel 2.1 Kit

Thermo Scientific Canine Genotypes Panel 2.1 is a reagents kit for canine parentage testings and identification using Short Tandem RepeatWeitere Informationen
Have Questions?
KatalognummerAnzahl Reaktionen
F864S100 Reaktionen
Katalognummer F864S
Preis (EUR)
2.375,00
Each
Anzahl Reaktionen:
100 Reaktionen
Preis (EUR)
2.375,00
Each
Thermo Scientific Canine Genotypes Panel 2.1 is a reagents kit for canine parentage testings and identification using Short Tandem Repeat (STR) DNA loci. Due to their high level of informativeness and Mendelian inheritance, microsatellites have become the markers of choice for parentage testing, identification and forensics.

The Canine Genotypes Panel 2.1 provides all the necessary reagents for amplification of the 19 microsatellite loci. The kit contains master mix and primer mix allows co-amplification of the microsatellites in a single multiplex PCR reaction. Canine control DNA is provided to verify acceptable PCR and electrophoresis conditions. Reagents and protocols of the Canine Genotypes Panel 2.1 kit have been optimized to deliver similar peak sizes (amplification yields) for each loci. Allele callings obtained with this kit represent the true alleles of an individual, instead of 'plus-A peaks' or 'split peaks' typically interpreted when using e.g. a conventional Taq DNA polymerase. The kit employs Phusion Hot Start DNA Polymerase that has a proofreading activity and eliminates the 'plus-A peaks'. One primer from each primer pair is end-labelled with a fluorescent dye. After PCR, the fragments are separated and detected in a single electrophoresis injection, using an automated capillary electrophoresis instrument.

Canine Genotypes Panel 2.1 is a kit for canine forensics, identification, parentage and breed testing. It encompasses the following 19 loci: PEZ02, PEZ17, FH2017, FH2309, PEZ05, FH2001, FH2328, FH2004, FH2361, PEZ21, FH2054, FH3377, FH2107, FH2088, vWF.X, FH2010, PEZ16 FH3313 and ZFX/Y (sex determining locus). These markers underwent a large developmental and validation study and were standardized for use in forensics (see references).

Highlights

• Single multiplex PCR of 19 STR loci
• Master mix and Primer mix provided for easy pipetting
• Control DNA provided for verification of PCR and electrophoresis conditions
• Allele calls represents true alleles (no plus-A peaks)

Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.

Specifications
NachweisverfahrenCapillary Electrophoresis
Zur Verwendung mit (Anwendung)PCR
Anzahl Reaktionen100 Reaktionen
ProdukttypCanine Genotypes Panel 2.1 Kit
ZielCanine Genotyping and Parentage Testing, Canine forensics
VerfahrenSTR (Short Tandem Repeat) Analysis
Unit SizeEach
Inhalt und Lagerung
Contains:
• 1.1 mL Canine Genotypes Panel 2.1 Master Mix
• 1.1 mL Canine Genotypes Panel 2.1 Primer Mix
• 30 μL Canine Genotypes 2.1 Control DNA

Store at -20°C. Primer Mix should be protected from light at all the times.

Häufig gestellte Fragen (FAQ)

Can additional markers be added to Thermo Scientific Animal Genotyping kits?

No, extra markers cannot be added to any of the Animal Genotyping kits. All primer mixes are optimized to work in a multiplex PCR. Addition of one or more primers would require optimization of the primer concentrations and the PCR protocol.

Are primers from Thermo Scientific Animal Genotyping kits available separately for specific loci?

We do not provide primers for specific locii separately. They come only in a multiplex format. If one or several loci did not amplify from the sample, faint signals can be a result of low DNA amounts or impurities. Some loci may be more sensitive towards purity of DNA used and may vary from sample to sample. On rare occasions, an animal can have a deletion in the marker region.

Do Thermo Scientific Animal Genotyping kits include an allelic ladder?

No, none of the genotyping kits contain an allelic ladder. Animal STR loci are highly polymorphic. Each allele has too many variants to make a universal allelic ladder.