FluoSpheres™ NeutrAvidin™-Labeled Microspheres, 0.04 μm, yellow-green fluorescent (505/515), 1% solids
Invitrogen™

FluoSpheres™ NeutrAvidin™-Labeled Microspheres, 0.04 μm, yellow-green fluorescent (505/515), 1% solids

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed fromRead more
Have Questions?
Catalog NumberQuantity
F87710.4 mL
Catalog number F8771
Price (USD)
628.00
Each
Quantity:
0.4 mL
Price (USD)
628.00
Each
Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Our Molecular Probes™ FluoSpheres™ beads are manufactured using high-quality, ultraclean polystyrene and are loaded with a variety of our proprietary dyes to create intensely fluorescent beads that typically show little or no photobleaching, even when excited with the intense illumination required for fluorescence microscopy. Different surface modifications are available to facilitate the coupling of various molecules and proteins to the surface of the bead.

FluoSpheres™ Microsphere Specifications

• Label (Ex/Em): Yellow-green fluorescent (505 / 515)
• Nominal bead diameter: 0.04 μm
• Surface modification: NeutrAvidin™
• Solids: 1%

Characteristics of the Various FluoSpheres™ Coupling Surfaces
• Carboxylate-modified FluoSpheres™ beads have a high density of pendent carboxylic acids on their surface, making them suitable for covalent coupling of proteins and other amine-containing biomolecules using water-soluble carbodiimide reagents such as EDAC
• Sulfate FluoSpheres™ beads are relatively hydrophobic particles that will passively and nearly irreversibly adsorb almost any protein, including albumin, IgG, avidin, and streptavidin
• Aldehyde-sulfate FluoSpheres™ beads have surface aldehyde groups added, designed to react with proteins and other amines under very mild conditions.
• Amine-modified FluoSpheres™ beads can be coupled to a wide variety of amine-reactive molecules, including the succinimidyl esters and isothiocyanates of haptens and drugs or the carboxylic acids of proteins, using a water-soluble carbodiimide

Key Applications of Microspheres
• Instrument calibration (flow cytometry, microscopy, HTS, HCS)
• Flow testing (microfluidics, blood flow, water flow, and air flow)
• Cell biology tracers (cell differentiation and cell tracing)
• Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

Choices for FluoSpheres™ Fluorescent Microspheres
Among our complete offering of fluorescent microspheres products, you’ll find beads with these variations:
• Ten fluorescent colors
• Ten nominal bead diameters: 0.02 μm, 0.04 μm, 0.1 μm, 0.2 μm, 0.5 μm, 1.0 μm, 2.0 μm, 4.0 μm, 10.0 μm, and 15.0 μm
• Four surface modifications for protein coupling: carboxylate, sulfate, aldehyde-sulfate, amine
• Microspheres that are additionally precoupled with streptavidin, NeutrAvidin, biotin, europium, and platinum

Choices for Unstained Microspheres
We also offer hundreds of choices for UltraClean™ surfactant-free microspheres for research and commercial applications.

We’ll Make a Custom Microsphere Product for You
We will prepare custom orders upon request. For example, FluoSpheres™ beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineFLUOSPHERES
Quantity0.4 mL
Shipping ConditionRoom Temperature
Surface ModificationOther Modification or Label
ColorYellow-Green
Diameter (Metric)0.04 μm
MaterialPolystyrene
Product TypeMicrosphere
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

What is the warranty for FluoSpheres microspheres?

The warranty period for FluoSpheres microspheres is 1-year from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

After washing and centrifugation, there was only a very small pellet left of my microsphere beads and the solution was transparent. Why is this?

Centrifugation is not an effective way to collect smaller microspheres; many particles remain in the solution even if you can visualize a small pellet. For beads less than 1 µm in diameter, we recommend washing by either:

Cross-flow filtration, as these particles have a very high compression modulus and can withstand high g-forces without risk of harm or dialysis with a 500 kDa MWCO
Note: Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I've had my microspheres for over a year, and I'm wondering if they're still good to use. What are some good ways to check their functionality?

Bacterial contamination is the most common cause of microspheres becoming unusable. Many of our particles are supplied with a low level of sodium azide to prevent bacterial contamination, but sometimes this can still occur. Bacterial contamination is best assessed by plating on appropriate growth medium and checking the plates after 72 hr.

Find additional tips, troubleshooting help, and resources within ourMicrospheres Support Center.

I accidentally froze my microspheres; can I still use them?

Even brief freezing can cause irreversible aggregation and potential distortion of the bead shape. You should not use these microspheres.

Find additional tips, troubleshooting help, and resources within our Microspheres Support Center.

My protein-coated microspheres appear to be non-specifically binding in my experimental system. Do you have a product that can help reduce these non-specific interactions?

Non-specific binding can often be relieved by a blocking solution, but microspheres seem to require a stronger blocking solution than those most commonly commercially available. Hence, we've developed the BlockAid Blocking Solution (Cat. No. B10710). This reagent is a protein-based blocking solution designed for use with FluoSpheres microspheres and TransFluoSpheres microspheres conjugated to biotin, streptavidin, NeutrAvidin biotin-binding protein, or other proteins. The BlockAid Blocking Solution has proven useful for reducing the nonspecific binding of protein-coated or other macromolecule-coated microspheres in a wide variety of flow cytometry, microscopy, and microarray applications.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (3)

Citations & References
Abstract
Polymer support for exonucleolytic sequencing.
Authors:Hinz M, Gura S, Nitzan B, Margel S, Seliger H
Journal:J Biotechnol
PubMed ID:11257537
'Different kinds of particles were investigated for their potential use as supports for exonucleolytic sequence analysis. Composite beads composed of an unreactive polystyrene "core" and a "shell" of functionalized silica nanoparticles were found to best fulfill the various prerequisites. The biotin/streptavidin system was used for attachment of DNA to composite ... More
Comparison of detection and signal amplification methods for DNA microarrays.
Authors:Vora GJ, Meador CE, Anderson GP, Taitt CR,
Journal:Mol Cell Probes
PubMed ID:18675897
One of the factors limiting the use of DNA microarray technology for the detection of pathogenic organisms from clinical and environmental matrices has been inadequate assay sensitivity. To assess the effectiveness of post-hybridization secondary detection steps to enhance the sensitivity of DNA microarray-based pathogen detection, we evaluated a panel of ... More
The strategy of T cell antigen-presenting cell encounter in antigen-draining lymph nodes revealed by imaging of initial T cell activation.
Authors:Bajénoff M, Granjeaud S, Guerder S
Journal:J Exp Med
PubMed ID:12953093
The development of an immune response critically relies on the encounter of rare antigen (Ag)-specific T cells with dendritic cells (DCs) presenting the relevant Ag. How two rare cells find each other in the midst of irrelevant other cells in lymph nodes (LNs) is unknown. Here we show that initial ... More