FluoSpheres™ Polystyrene Microspheres, 10 μm, blue fluorescent (365/415), for blood flow determination
Invitrogen™

FluoSpheres™ Polystyrene Microspheres, 10 μm, blue fluorescent (365/415), for blood flow determination

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed fromRead more
Have Questions?
Catalog NumberQuantity
F882910 mL
Catalog number F8829
Price (USD)
572.00
Each
Quantity:
10 mL
Request bulk or custom format
Price (USD)
572.00
Each
Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Our Molecular Probes™ FluoSpheres™ beads are manufactured using high-quality, ultraclean polystyrene and are loaded with a variety of our proprietary dyes to create intensely fluorescent beads that typically show little or no photobleaching, even when excited with the intense illumination required for fluorescence microscopy.

FluoSpheres™ microspheres in the 10–15 μm size range were designed specifically for regional blood flow studies in tissues and organs. Typically for these types of studies, the microspheres are injected at desired locations in the circulatory system and eventually lodge in the capillaries, where they can be extracted and the fluorescence quantified. The use of fluorescent microspheres eliminates the radiation hazards and disposal hurdles associated with radiolabeled microspheres. Multiple (up to seven or more with good spectral correction) simultaneously circulating FluoSpheres™ microspheres can be discriminated. Microspheres that have been perfused or injected into tissues and organs retain their fluorescence following histological serial sectioning. Our FluoSpheres™ microspheres for blood flow determination, which are available in eleven distinguishable fluorescent colors, are also compatible with blood flow analyzer systems that perform automated extraction and analysis.

FluoSpheres™ Microsphere Specifications:

• Label (Ex/Em): Blue fluorescent (365/415)
• Nominal bead diameter: 10 μm

Choices for FluoSpheres™ Fluorescent Microspheres
In addition to microspheres for blood flow studies, investigate our complete offering of fluorescent microspheres products. Among those products you’ll find beads with these variations:
• Ten fluorescent colors
• Ten nominal bead diameters: 0.02 μm, 0.04 μm, 0.1 μm, 0.2 μm, 0.5 μm, 1.0 μm, 2.0 μm, 4.0 μm, 10.0 μm, and 15.0 μm
• Four surface modifications for protein coupling: carboxylate, sulfate, aldehyde-sulfate, amine
• Microspheres that are additionally precoupled with streptavidin, NeutrAvidin, biotin, europium, and platinum

Choices for Unstained Microspheres
We also offer hundreds of choices for UltraClean™ surfactant-free microspheres for research and commercial applications.

We’ll Make a Custom Microsphere Product for You
We will prepare custom orders upon request. For example, FluoSpheres™ beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration3.6 x 106 beads/mL
Product LineFLUOSPHERES
Quantity10 mL
Shipping ConditionRoom Temperature
ColorBlue
Diameter (Metric)10 μm
MaterialPolystyrene
Product TypeMicrosphere
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

I want to use fluorescent microspheres for blood tracing studies. What functional group should be on the surface?

We offer FluoSpheres fluorescent microspheres for tracer studies that are 1, 10 and 15 µm in diameter, with various wavelengths. If those sizes or available colors are not optimal for your application, then we recommend carboxylate-modified microspheres, which are available in a wide range of sizes and colors.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to use fluorescent microspheres for blood tracing studies. What functional group should be on the surface of the beads?

We offer FluoSpheres fluorescent microspheres that are available in 10 and 15 µm diameters, with various wavelengths, for tracer studies. If those sizes or available colors are not optimal for your system, then we recommend the carboxylate-modified microspheres, which are available in a wide range of sizes and colors.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the warranty for FluoSpheres microspheres?

The warranty period for FluoSpheres microspheres is 1-year from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

After washing and centrifugation, there was only a very small pellet left of my microsphere beads and the solution was transparent. Why is this?

Centrifugation is not an effective way to collect smaller microspheres; many particles remain in the solution even if you can visualize a small pellet. For beads less than 1 µm in diameter, we recommend washing by either:

Cross-flow filtration, as these particles have a very high compression modulus and can withstand high g-forces without risk of harm or dialysis with a 500 kDa MWCO
Note: Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I've had my microspheres for over a year, and I'm wondering if they're still good to use. What are some good ways to check their functionality?

Bacterial contamination is the most common cause of microspheres becoming unusable. Many of our particles are supplied with a low level of sodium azide to prevent bacterial contamination, but sometimes this can still occur. Bacterial contamination is best assessed by plating on appropriate growth medium and checking the plates after 72 hr.

Find additional tips, troubleshooting help, and resources within ourMicrospheres Support Center.

Citations & References (3)

Citations & References
Abstract
Validation of fluorescent-labeled microspheres for measurement of regional organ perfusion.
Authors:Glenny RW, Bernard S, Brinkley M
Journal:J Appl Physiol
PubMed ID:8335595
'Estimations of dog lung, pig heart, and pig kidney regional perfusion by use of fluorescent-labeled microspheres were compared with measurements obtained with standard radiolabeled microspheres. Pairs of radio- and fluorescent-labeled microspheres (15 microns diam, 6 colors) were injected into a central vein of a supine anesthetized dog and the left ... More
Blood flow measurements by the reference sample method with microsphere injection in to the aorta: an accurate and easy approach.
Authors:Sanchez S, Bandi JC, Mastai R
Journal:Proc Soc Exp Biol Med
PubMed ID:1615013
'The validity of hemodynamic measurements by the reference sample method with microspheres injection into the aorta, via a carotid artery catheter, was evaluated in rats and compared with the results obtained after left ventricle injection. In the aorta injection group, a good mix of microspheres was observed in 83% of ... More
Fluorescent microspheres to measure organ perfusion: validation of a simplified sample processing technique.
Authors:Van Oosterhout MF, Willigers HM, Reneman RS, Prinzen FW
Journal:Am J Physiol
PubMed ID:7653638
A disadvantage of nonradioactive microsphere techniques is that the processing of samples is time-consuming and complex. We developed and validated a simplified processing method for the fluorescent microsphere (FM) technique. In seven anesthetized dogs with coronary artery stenosis up to six different FM and five different radioactivity labeled microspheres (RM) ... More