FluoSpheres™ Sulfate-Modified Microspheres, 0.02–4.0 μm
Invitrogen™

FluoSpheres™ Sulfate-Modified Microspheres, 0.02–4.0 μm

Perform flow cytometry, microscopy, HTS, and HCS with our FluoSpheres Sulfate-Modified Microspheres, featuring different bead diameters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Color
F88594 μmYellow-Green
F8851
also known as F-8851
1 μmRed
F88584 μmRed
F8845
also known as F-8845
0.02 μmYellow-Green
F88480.20 μmYellow-Green
F88521 μmYellow-Green
F88532 μmYellow-Green
Catalog number F8859
Price (USD)
397.65
Online exclusive
530.00
Save 132.35
Each
Diameter (Metric):
4 μm
Color:
Yellow-Green
Price (USD)
397.65
Online exclusive
530.00
Save 132.35
Each
Easily perform flow cytometry, microscopy, HTS, HCS, and other laboratory applications using our extensive selection of FluoSpheres Sulfate-Modified Microspheres. These microsphere beads can be used in passive adsorption or active, covalent coupling of proteins, nucleic acids, and biomolecules for particle capture applications.

FluoSpheres Sulfate-Modified Microspheres are loaded with our proprietary fluorescent dyes, making them the brightest microspheres available. Because the fluorescent dye molecules are contained inside each polystyrene microsphere instead of only on the surface, the dye molecules are shielded from many of the environmental effects that cause quenching or photobleaching of exposed fluorophores. The stability, uniformity, and reproducibility of FluoSpheres microspheres, as well as their extensive color selection, make them the preferred tools for research and diagnostic assays that use fluorescence.

FluoSpheres Sulfate-Modified Microspheres are relatively hydrophobic particles that will passively and nearly irreversibly adsorb almost any protein, including albumin, IgG, avidin, and streptavidin.

Key applications of FluoSpheres Sulfate-Modified Microspheres
• Instrument calibration (flow cytometry, microscopy, HTS, HCS)
• Flow testing (microfluidics, blood flow, water flow, and air flow)
• Cell biology tracers (cell differentiation and cell tracing)
• Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

FluoSpheres sulfate-modified microsphere specifications for F8845, F8848, F8852, F8853, and F8859:
• Label (excitation/emission): yellow-green (505/515)
• Nominal bead diameters: 0.02, 0.2, 1.0, 2.0, and 4.0 μm
• Coupling surface: sulfate
• Solids: 2%

FluoSpheres sulfate-modified microsphere specifications for F8851 and F8858:
• Red (580/605)
• Nominal bead diameters: 1.0, 4.0 μm
• Coupling surface: sulfate
• Solids: 2%

FluoSpheres fluorescent microspheres are potentially more sensitive than colorimetric methods in most, if not all, major microsphere-based diagnostic test systems presently in use, including microsphere-agglutination tests, filter separation tests, particle-capture ELISA methods, and two-particle sandwich techniques.

FluoSphere Sulfate-Modified Microspheres (also called latex beads or latex particles) are manufactured using high-quality, ultra-clean polystyrene and are loaded with a variety of our proprietary dyes to create intensely fluorescent beads that typically show little or no photobleaching, even when excited with the intense illumination required for fluorescence microscopy. Every possible precaution is taken throughout the manufacturing process to ensure that the microparticles are kept free of contaminating agents. The final product is sold as a suspension in ultrapure water, in most cases containing 2 mM azide or 0.02% thimerosal as a preservative.

Other surface modifications are available to facilitate the coupling of various molecules and proteins to the surface of the bead, including amine- and carboxylate-modified FluoSphere microspheres.

Choices for unstained microspheres
We also offer hundreds of choices for ultra-clean surfactant-free microspheres for research and commercial applications.

Custom microspheres
We prepare custom microspheres upon request. For example, FluoSpheres Sulfate-Modified microsphere beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineFLUOSPHERES
Quantity2 mL
Shipping ConditionRoom Temperature
Surface ModificationSulfate
Volume (Metric)2 mL
ColorYellow-Green
Diameter (Metric)4 μm
For Use With (Application)Cell Analysis
MaterialPolystyrene
Product TypeMicrosphere
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

I have some FluoSpheres polystyrene microspheres, with 20 nm diameter. They are aggregating a lot. What can I do about it?

The smaller the microspheres, the greater the propensity to aggregate. But the aggregation is not irreversible. Sonicate in a bath sonicator or vortex to disperse, just prior to use. You can also add a small concentration of Tween-20 or Triton X-100 (unless you are using them in a live-cell system).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I sonicated my 2.0 µm carboxylate-modified microspheres, as recommended, but saw foaming (bubbles) on top of the solution. Should I be concerned?

Use of a bath sonicator is recommended to help break up any aggregated microspheres. The foaming is from Tween-20, which is in the stock solution to help prevent aggregation. It is normal and expected to see bubbles from this. Do not use a probe sonicator, which would cause damage to the microspheres (as well as much more bubbling).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the warranty for FluoSpheres microspheres?

The warranty period for FluoSpheres microspheres is 1-year from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

After washing and centrifugation, there was only a very small pellet left of my microsphere beads and the solution was transparent. Why is this?

Centrifugation is not an effective way to collect smaller microspheres; many particles remain in the solution even if you can visualize a small pellet. For beads less than 1 µm in diameter, we recommend washing by either:

Cross-flow filtration, as these particles have a very high compression modulus and can withstand high g-forces without risk of harm or dialysis with a 500 kDa MWCO
Note: Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I've had my microspheres for over a year, and I'm wondering if they're still good to use. What are some good ways to check their functionality?

Bacterial contamination is the most common cause of microspheres becoming unusable. Many of our particles are supplied with a low level of sodium azide to prevent bacterial contamination, but sometimes this can still occur. Bacterial contamination is best assessed by plating on appropriate growth medium and checking the plates after 72 hr.

Find additional tips, troubleshooting help, and resources within ourMicrospheres Support Center.

Citations & References (3)

Citations & References
Abstract
Aspirin inhibits in vitro maturation and in vivo immunostimulatory function of murine myeloid dendritic cells.
Authors:Hackstein H, Morelli AE, Larregina AT, Ganster RW, Papworth GD, Logar AJ, Watkins SC, Falo LD, Thomson AW
Journal:J Immunol
PubMed ID:11390449
'Aspirin is the most commonly used analgesic and antiinflammatory agent. In this study, at physiological concentrations, it profoundly inhibited CD40, CD80, CD86, and MHC class II expression on murine, GM-CSF + IL-4 stimulated, bone marrow-derived myeloid dendritic cells (DC). CD11c and MHC class I expression were unaffected. The inhibitory action ... More
Improved protocol for a microsphere-adhesion assay on living tissue slices.
Authors:Förster E, Kaltschmidt C
Journal:Biotechniques
PubMed ID:10090988
Here, we describe a simple microsphere-adhesion assay to characterize adhesive cues on living tissue slices, which we use to study pattern formation in neural tissue. This assay was developed by modifying a cell-adhesion assay on living tissue slices. We replaced dissociated cells by fluorescent microspheres and then coated the microspheres ... More
Disialosyl galactosylgloboside as an adhesion molecule expressed on renal cell carcinoma and its relationship to metastatic potential.
Authors:Satoh M, Handa K, Saito S, Tokuyama S, Ito A, Miyao N, Orikasa S, Hakomori S
Journal:Cancer Res
PubMed ID:8620516
Aberrant glycosylation expressed in specific types of human cancer may define stage, direction, and fate of tumor progression. Well-studied examples are expression of sialosyl-Lewis(x) or sialosyl-Lewis(a) in colorectal carcinoma and histo-blood group A and H/Le(y) in lung cancer. In renal cell carcinoma (RCC), expression of sialosyl-Lewis(x) has no correlation with ... More