Product Image
Thermo Scientific™

Bovine Genotypes Panel 3.1 Kit

Thermo Scientific Bovine Genotypes Panel 3.1 Kit is offered for cattle parentage testing and identification using Short Tandem Repeat (STR)Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
F900L500 Reactions
F900S100 Reactions
Catalog number F900L
Price (USD)
12,610.00
Each
No. of Reactions:
500 Reactions
Price (USD)
12,610.00
Each
Thermo Scientific Bovine Genotypes Panel 3.1 Kit is offered for cattle parentage testing and identification using Short Tandem Repeat (STR) DNA loci.

The Bovine Genotypes Panel 3.1 allows co-amplification of the 18 microsatellites in a single multiplex PCR reaction. One primer from each primer pair is end-labeled with a fluorescent dye. After PCR, the fragments are separated and detected in a single electrophoresis injection, using an automated electrophoresis instrument (see Figure 2). In addition, the kit includes bovine control DNA for verification of acceptable PCR and electrophoresis conditions. Reagents and protocols of the Bovine Genotypes Panel 3.1 have been optimized to deliver similar peak sizes (amplification yields) for each loci. Allele callings obtained with this kit represent the true alleles of an individual, instead of 'plus-A peaks' or 'split peaks' typically interpreted when using e.g. a conventional Taq DNA polymerase. The kit employs Phusion Hot Start DNA Polymerase that has a proofreading activity and eliminates the 'plus-A peaks'.

Thermo Scientific Bovine Genotypes Panel 3.1 encompasses all the 12 STR loci recommended by the International Society for Animal Genetics (ISAG) for routine use in parentage testing and identification, including TGLA227, BM2113, TGLA53, ETH10, SPS115, TGLA126, TGLA122, INRA23, ETH3, ETH225, BM1824, and BM1818. In addition, the kit includes the following six microsatellites, which are among the list of loci recommended by the Food and Agriculture Organization of the United Nations (FAO) for genetic studies of domestic animals: SPS113, RM067, CSRM60, MGTG4B, CSSM66, and ILSTS006. Bovine Genotypes Panel 3.1 is a combination of Panels 1.2 and 2.2, thus enabling the analysis of both panels in a single reaction.

Highlights

• Various kit choices that include STR loci recommended by ISAG
• Single multiplex reaction - less pipetting steps
• Optimized reagents
• Control DNA provided for verification of PCR and electrophoresis conditions
• Allele calling represents true alleles (no plus-A peaks)

This product has been developed in collaboration with Maxxam Analytics Inc.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodCapillary Electrophoresis
For Use With (Application)PCR
No. of Reactions500 Reactions
Product TypeBovine Genotypes Panel 3.1 Kit
Quantity500 reactions
TargetBovine Genotyping and Parentage Testing
TechniqueSTR (Short Tandem Repeat) Analysis
Unit SizeEach
Contents & Storage
Contains:
• 5 x 1.1 mL Bovine Genotypes Master Mix
• 5 x 1.1 mL Bovine Genotypes Panel 3.1 Primer Mix
• 150 μL Bovine Genotypes Control DNA001

Store at -20°C. Primer Mix should be protected from light at all the times.

Frequently asked questions (FAQs)

Can additional markers be added to Thermo Scientific Animal Genotyping kits?

No, extra markers cannot be added to any of the Animal Genotyping kits. All primer mixes are optimized to work in a multiplex PCR. Addition of one or more primers would require optimization of the primer concentrations and the PCR protocol.

Are primers from Thermo Scientific Animal Genotyping kits available separately for specific loci?

We do not provide primers for specific locii separately. They come only in a multiplex format. If one or several loci did not amplify from the sample, faint signals can be a result of low DNA amounts or impurities. Some loci may be more sensitive towards purity of DNA used and may vary from sample to sample. On rare occasions, an animal can have a deletion in the marker region.

Do Thermo Scientific Animal Genotyping kits include an allelic ladder?

No, none of the genotyping kits contain an allelic ladder. Animal STR loci are highly polymorphic. Each allele has too many variants to make a universal allelic ladder.