Sterile PETG Media Bottles, 1000mL - FAQs
View additional product information for Sterile PETG Media Bottles - FAQs (FBMB1000)
1 product FAQs found
What is the minimum temperature at which I can use PETG media bottles?
We have tested our PETG bottles down to -70 degrees C. Please refer to the Thermo Scientific Nalgene PETG bottle performance at -70 degrees C Application Note.