I am using counting beads to count cells, but I cannot find the beads on my scatter plots. What do I do?
The first thing to do is check your threshold and see if it is set on forward scatter. If so, the beads are probably being excluded by the threshold. Reducing the threshold setting should reveal your beads.