Product Image
Invitrogen™

Fetal Hemoglobin Control Kit (FETALtrol™)

FETALtrol™ is a tri-level, assayed human blood control designed to help document and monitor values obtained from test methods used to determine fetal RBCs in maternal blood samples.
Have Questions?
Catalog NumberQuantity
FH10150 Tests
Catalog number FH101
Price (USD)
572.00
Each
Quantity:
50 Tests
Price (USD)
572.00
Each

FETALtrol™ is a tri-level, assayed human blood control designed to help document and monitor values obtained from test methods used to determine fetal RBCs in maternal blood samples. It monitors preparation techniques, stains, reagents, and methods of data analysis for quantitative and qualitative tests. FETALtrol™ is reagent comprising D-antigen (Rho) negative adult human erythrocytes supplemented with D-antigen (Rho) positive human cord blood erythrocytes.

Application
The laboratory determination of the level of fetal cells in maternal circulation remains an important support in the obstetrical management of women with suspected uterine trauma and in the proper dose administration of Rh immune globulin. The Trillium FETALtrol™ kit provides a means of monitoring preparation techniques, stains, reagents, and methods of data analysis for quantitative and qualitative tests.

Additional Information
Use the Spectraviewer to help you plan your experiments. Additional general information about Life Technologies flow cytometry products and support is also available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications
For Use With (Application)Flow Cytometry, Immunohistochemistry
For Use With (Equipment)Microscope (KBB assay-compatible), Flow Cytometer
FormatTube
Quantity50 Tests
TypeFetal Hemoglobin Control Kit
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).

Citations & References (6)

Citations & References
Abstract
Detection of fetal red cells in fetomaternal hemorrhage using a fetal hemoglobin monoclonal antibody by flow cytometry.
Authors:Davis BH, Olsen S, Bigelow NC, Chen JC
Journal:Transfusion
PubMed ID:9709783
'The laboratory determination of the level of fetal cells in maternal circulation remains an important support in the obstetrical management of women with suspected uterine trauma and in the proper dose administration of anti-D for prevention of Rh hemolytic disease of the newborn. Limitations in the sensitivity and precision of ... More
[Demonstration of fetal hemoglobin in erythrocytes of a blood smear].
Authors:KLEIHAUER E, BRAUN H, BETKE K
Journal:Klin Wochenschr
PubMed ID:13450287
Multicenter clinical experience with flow cytometric method for fetomaternal hemorrhage detection.
Authors:Chen JC, Davis BH, Wood B, Warzynski MJ
Journal:Cytometry
PubMed ID:12497589
Enumeration of fetal red blood cells (RBCs) is important in the management of fetomaternal hemorrhage (FMH), particularly in situations of Rh incompatibility.
Occupational exposure to bloodborne pathogens; needlestick and other sharps injuries; final rule. Occupational Safety and Health Administration (OSHA), Department of Labor. Final rule; request for comment on the Information Collection (Paperwork) Requirements.
Authors:
Journal:Fed Regist
PubMed ID:11503775
The Occupational Safety and Health Administration is revising the Bloodborne Pathogens standard in conformance with the requirements of the Needlestick Safety and Prevention Act. This Act directs OSHA to revise the Bloodborne Pathogens standard to include new examples in the definition of engineering controls along with two new definitions; to ... More
Occupational exposure to bloodborne pathogens--OSHA. Final rule.
Authors:
Journal:Fed Regist
PubMed ID:10115865
The Occupational Safety and Health Administration hereby promulgates a standard under section 6(b) of the Occupational Safety and Health Act of 1970 (the Act), 29 U.S.C. 655 to eliminate or minimize occupational exposure to Hepatitis B Virus (HBV), Human Immunodeficiency Virus (HIV) and other bloodborne pathogens. Based on a review ... More