Beta Gauge Continuous Ambient Particulate Monitor, FH62C14
Thermo Scientific™

Beta Gauge Continuous Ambient Particulate Monitor, FH62C14

Continuously measure the mass concentration of particulates using beta attenuation with the Thermo Scientific™ Beta Gauge Continuous Ambient Particulate Monitor.
Have Questions?
Número de catálogoDescripción
FH62C14FH62C14 Series Continuous Ambient Particulate Monitor
Número de catálogo FH62C14
Precio (MXN)
-
Descripción:
FH62C14 Series Continuous Ambient Particulate Monitor

Continuously measure the mass concentration of particulates using beta attenuation with the Thermo Scientific™ Beta Gauge Continuous Ambient Particulate Monitor, FH 62 C14. This continuous (non-step wise) ambient monitor measures airborne particulates with long-term precision.

The Thermo Scientific Continuous Ambient Particulate Monitor measures PM-10, PM-2.5 or PM-1 mass concentrations, and consists of a C14 source and detector in a compact 19-inch rack-mountable enclosure. The FH62C14 automatically advances the filter tape based upon a user-defined frequency or particulate loading.

Key Features:

  • Designed to meet U.S. and International Particulate Monitoring Regulations
  • Simultaneous sample collection and measurement on a single sample spot located between the C14 source and the proportional detector
  • Dynamic filter loading is measured continuously by the attenuation of the C14 source beta rays while ambient particulate matter is being deposited onto a filter tape sample spot.
  • Filter spot does not need to be moved from the sample position to the detector position for zero and mass determination

Additional Information:

  • Monitor permits control and data exchange over two serial interfaces
  • Store half-hour average concentrations over a whole year
  • Features user-selectable reporting of mass concentration based on standard or actual flow rate
  • Calibration of all sensors is processor-controlled
  • Measurement Cycle: Single filter spot in position for 24 hours (default); user-selectable 30 minutes to 24 hours
  • Data Averages/Values Displayed: Each full 1/2 and 24-hour values automatically stored; each full, 1/2, 1, 3, and 24-hour

Includes:

  • Temperature sensors
  • Communication cables
  • Consumables for average first year operation (ambient)

Compatible with:

Complete Outdoor Enclosure, Streamline PRO

Recommended for:

  • Air quality monitoring networks, including background sites
  • Special studies and supersites
  • Routine input for air quality index
  • In and around industrial and material handling facilities
  • Remediation projects (Superfund, hazardous waste)
  • Indoor air, exposure chamber, and industrial hygiene measurements
Especificaciones
Caudal0 to 20 L/min.
Método de mediciónContinuous and Simultaneous Particulate Collection through Beta Ray Attenuation
Temperatura de funcionamiento (imperial)-22°F to +140°F
Temperatura de funcionamiento (métrico)-30°C to +60°C
Certificaciones/conformidadUSEPA Equivalent PM-10 Method: EQPM-1102-150CARB California Approved Sampler (CAS) for PM-10 and PM-2.5
Fondo (imperial)13 in.
Fondo (métrico)330 mm
DescripciónFH62C14 Series Continuous Ambient Particulate Monitor
Frecuencia50/60 Hz
Altura (imperial)12.25 in.
Altura (métrico)311 cm
IncluyeTemperature sensors, communication cables, consumables for average first year operation (ambient)
Intervalo de medición0 to 5000 μg/m3 or 0 to 10,00 0 μg/m3
SalidasOutput: Serial interface RS-232 Analog output; 0-10V 4-20mA output of concentration (µg/cu. m)
Precisión2 μg/m3 (24-hour)
Tensión100/240 V
Vataje330 w (Instrument); 100 w (Pump)
Peso (imperial)50 lb. (Instrument)/18 lb. (Pump)
Peso (métrico)18 kg (Instrument)/8.2 kg (Pump)
Anchura (imperial)19 in.
Anchura (métrico)483 mm
Unit SizeEach