PackEye Radiation Detection Backpack
Actual product may vary
Thermo Scientific™
PackEye Radiation Detection Backpack
Provide your survey teams with a tool for effectively addressing the problems of orphaned sources, radiological contamination, and maliciously introduced sources.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호설명
FHT1377FHT PackEye Radiation detection backpack
PACKEYEDS33 year extended depot service warranty
카탈로그 번호 FHT1377
제품 가격(KRW)
-
설명:
FHT PackEye Radiation detection backpack
Provide your survey teams with a tool for effectively addressing the problems of orphaned sources, radiological contamination, and maliciously introduced sources with Thermo Scientific™ PackEye Radiation Detection Backpack. It locates and detects gamma-emitting radioactive sources in large areas very rapidly, but unobtrusively.
  • Fast detection and locating of radioactive sources via high sensitivity plastic detector
  • Easy nuisance alarm management by immediate discrimination between natural (NORM, natural background changes) and artificial radiation via Natural Background Rejection algorithm (NBR) 
  • Always maximum sensitivity through automatic adjustment to background variations
  • High neutron detection sensitivity achieved with He-3 free detector technology
  • Gamma only version available
  • User friendly interface with simple LED level, alarm and status indication including NBR feedback (red and green)
  • Stealth mode using standard earphone muting audible level indication or alarm
  • Unchallenged light weight of 7.5 kg
  • Low power consumption - operation time 30hr (20hr with Bluetooth™ data transfer to optional PDA)
  • Can be tied into the Thermo Scientific™ ViewPoint™ Enterprise platform, allowing for a customizable remote monitoring solution which will provide real-time command and control data
The Backpack Includes:
  • 2 ea. flat Li-6 scintillation detectors with proprietary measurement technique, allowing minimal crosstalk while achieving a very low net alarm threshold level 
  • Highly sensitive NBR plastic scintillation detector with voltage divider and Photomultiplier
  • Low power controller (type FHT 681) with attached high capacity battery module and user friendly interface (LED based indicator unit)
  • Accessory case with connection cables to PC or notebook, spare battery pack, earphone, set of international power socket plugs for charger, car adapter as alternative power supply for the battery charger and rain cover
Recommended Accessories:
  • Thermo Scientific™ RadEye™ SPRD with belt holster: Spectroscopic Personal Radiation Detector (SPRD) for flexible source location, dose rate survey and nuclide identification in case of alarm.
  • PDA Set for convenient data display including GPS (connected to the PackEye Backpack via Bluetooth)
  • Transportation case with outside power socket
  • Tripod for transportation case allowing the PackEye Backpack to be used as transportable, temporarily stationary Gamma and Neutron monitor
사양
알람Artificial gamma alarm: Typically better than 20% of natural background
배터리 수명30 hr. (20 hr with PDA operation via Bluetooth)
배터리 종류Rechargeable NiMH, power pack (7.2 V)
깊이(영국식 단위)7 in.
깊이(미터법)18 cm
설명FHT PackEye Radiation detection backpack
에너지 범위20 keV to 3 MeV
높이(영국식 단위)22.8 in.
높이(미터법)580 mm
방사선 유형Gamma and Neutron (Gamma only available)
민감도Neutron ( 40 cps per n / s/cm2)
온도(영국식 단위) 범위-4°F to +122°F
온도(미터법) 범위-20°C to +50°C
중량(영국식 단위)16.5 lb.
중량(미터법)6 kg
폭(영국식 단위)11.8 in.
폭(미터법)300 mm
제품 유형Radiation Detection Backpack
Unit SizeEach