What is TransFix Cellular Antigen Stabilizing Reagent?
TransFix Cellular Antigen Stabilizing Reagent (Cat. Nos. FIX2, FIX20, and FIX100) is a whole blood preservative used for the stabilization of leukocytes and the preservation of antigenic sites for future cellular analysis. It enables simple batch-processing of multiple samples for optimal laboratory workflow. Additionally, blood that has been stabilized with TransFix reagent can be collected and transported to other sites for analysis. It is easy to use, cost-effective, proven in multiple publications, convenient, and flexible.